ABsolute QPCR Mix, SYBR Green, ROX
Product Image
Thermo Scientific™

ABsolute QPCR Mix, SYBR Green, ROX

**********************Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
AB1163A400 Reactions
AB1162B1,600 Reactions
Catalog number AB1163A
Price (INR)
66,631.00
Each
Add to cart
No. of Reactions:
400 Reactions
Price (INR)
66,631.00
Each
Add to cart
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of ABsolute QPCR Mix, SYBR Green, and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute qPCR SYBR Green Mixes are optimized for SYBR Green chemistry and contain all the components necessary to perform quantitative PCR, with the exception of template and primers. The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.

ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.

Highlights

• High Sensitivity & Specificity—The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
• Optimised buffer containing SYBR Green I
• Reproducibility—All master mixes undergo extensive QC to ensure batch to batch consistency
• Broad dynamic range

Applications

• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute QPCR Mix, no ROX
ABsolute QPCR Mix, low ROX
ABsolute QPCR Mix, ROX
ABsolute QPCR Capillary Mix
ABsolute QPCR Mix, SYBR Green, no ROX
ABsolute QPCR Mix, SYBR Green, low ROX
ABsolute QPCR Mix, SYBR Green, ROX
ABsolute QPCR Mix, SYBR Green, fluorescein
ABsolute QPCR Capillary Mix, SYBR Green
For Research Use Only. Not for use in diagnostic procedures.
Specifications
CompatibilityqPCR Cyclers
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
No. of Reactions400 Reactions
PolymeraseTaq DNA Polymerase
Product LineABsolute
Product TypeReal Time PCR SYBR Master Mix
Quantity400 reactions
Sample TypeDNA, dsDNA
Sufficient For400 Reactions of 25 μL Each
Volume5 mL
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression, Genotyping, siRNA Validation
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains:
• 1 x 5 mL ABsolute QPCR SYBR Green ROX Mix (2X)
• 100 μL MgCl2 (1M)
Sufficient for 400 x 25 μL reactions.

Store at -20°C