Thermo Scientific Chemicals

n-Dodecyl-beta-D-maltopyranoside, Thermo Scientific Chemicals

Catalog number: J66869.06
5 g, Each
Thermo Scientific Chemicals

n-Dodecyl-beta-D-maltopyranoside, Thermo Scientific Chemicals

Catalog number: J66869.06
5 g, Each
Quantity
Catalog number: J66869.06
also known as J66869-06
Price (JPY)

Chemical Identifiers

CAS
69227-93-6
IUPAC Name
(2R,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6R)-6-(dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C24H46O11
InChI Key
NLEBIOOXCVAHBD-QKMCSOCLSA-N
SMILES
CCCCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O
Optical Rotation
+45 to +49?, c=1 in water
Form
Powder
Appearance (Color)
White
Assay (HPLC)
≥ 99.0%

Description

N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

Solubility
Soluble in water at 10% w/v

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text