Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000454
          Assay Name: hsa-miR-130a
          miRBase Accession Number: MI0000448
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 57641198 - 57641286 [+] on Build GRCh38
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Species: Human, Mouse, Rat, Bovine, Chicken, Columba livia, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Guinea pig, Horse, Monodelphis domestica, Ophiophagus hannah, Ornithorhynchus anatinus, Otolemur garnettii, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Taeniopygia guttata, Tupaia chinensis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000454
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000034, mir-130

          Mature miRNA Sequence:

          CAGUGCAAUGUUAAAAGGGCAU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-130a-3p MIMAT0000425
          Mouse mmu-miR-130a-3p MIMAT0000141
          Rat rno-miR-130a-3p MIMAT0000836
          Bovine bta-miR-130a MIMAT0009223
          Chicken gga-miR-130c-3p MIMAT0007734
          Columba livia cli-miR-130a-3p MIMAT0038506
          View More Rows
          Cricetulus griseus cgr-miR-130a-3p MIMAT0023755
          Cyprinus carpio ccr-miR-130a MIMAT0026210
          Dasypus novemcinctus dno-miR-130a-3p MIMAT0047628
          Dog cfa-miR-130a MIMAT0006631
          Guinea pig cpo-miR-130a-3p MIMAT0046981
          Horse eca-miR-130a MIMAT0013047
          Monodelphis domestica mdo-miR-130c-3p MIMAT0028515
          Ophiophagus hannah oha-miR-130b-3p MIMAT0036690
          Ornithorhynchus anatinus oan-miR-130b-3p MIMAT0007159
          Otolemur garnettii oga-miR-130a MIMAT0049666
          Pan troglodytes ptr-miR-130a MIMAT0008025
          Pig ssc-miR-130a MIMAT0007758
          Pongo pygmaeus ppy-miR-130a MIMAT0015753
          Pteropus alecto pal-miR-130-3p MIMAT0039896
          Python bivittatus pbv-miR-130d-3p MIMAT0038932
          Taeniopygia guttata tgu-miR-130c-3p MIMAT0014581
          Tupaia chinensis tch-miR-130a-3p MIMAT0036577
          Xenopus tropicalis xtr-miR-130a MIMAT0003591
          Zebrafish dre-miR-130a MIMAT0001826

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-130a
          Stem-loop Accession # MI0000448
          Stem-loop Sequence
          UGCUGCUGGCCAGAGCUCUUUUCACAUUGUGCUACUGUCUGCACCUGUCACUAGCAGUGCAAUGUUAAAAGGGCAUUGGCCGUGUAGUG
          Chromosome Location Chr. 11 - 57641198 - 57641286 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000425
          miRBase ID: hsa-miR-130a-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: Chr. 11 - 57641198 - 57641286 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-130a
          Stem-loop Accession # MI0000156
          Stem-loop Sequence
          GAGCUCUUUUCACAUUGUGCUACUGUCUAACGUGUACCGAGCAGUGCAAUGUUAAAAGGGCAUC
          Chromosome Location Chr. 2 - 84741115 - 84741178 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000141
          miRBase ID: mmu-miR-130a-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: Chr. 2 - 84741115 - 84741178 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-130a
          Stem-loop Accession # MI0000903
          Stem-loop Sequence
          UGCUGCUGGCCGGAGCUCUUUUCACAUUGUGCUACUGUCUACACGUGUACCGAGCAGUGCAAUGUUAAAAGGGCAUCGGCCUUGUAGU
          Chromosome Location Chr. 3 - 72141037 - 72141124 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000836
          miRBase ID: rno-miR-130a-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: Chr. 3 - 72141037 - 72141124 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-130a
          Stem-loop Accession # MI0009730
          Stem-loop Sequence
          UGCUGCGGGCCGGAGCUCUUUUCACAUUGUGCUACUGUCUGCGCCUGUCACUAGCAGUGCAAUGUUAAAAGGGCAUUGGCUGUGCAGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009223
          miRBase ID: bta-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-130c
          Stem-loop Accession # MI0007560
          Stem-loop Sequence
          UUGAGCGGUGUCUGUCCAGCGCCCUUUUUAUGUUGUACUACUAGUGAUCAUACACAAAGCAGUGCAAUGUUAAAAGGGCAUUGGCCAGCAGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007734
          miRBase ID: gga-miR-130c-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-130a
          Stem-loop Accession # MI0029962
          Stem-loop Sequence
          CUGUCCAGUGCCCUUUUUAUGUUGUACUACUGGUGAUCAUGCACAAAAAGCAGUGCAAUGUUAAAAGGGCAUUGGCCAGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038506
          miRBase ID: cli-miR-130a-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-130a
          Stem-loop Accession # MI0020394
          Stem-loop Sequence
          GCUGGCCGGAGCUCUUUUCACAUUGUGCUACUGUCUUCACCUAUAACGAGCAGUGCAAUGUUAAAAGGGCAUCGGCCUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023755
          miRBase ID: cgr-miR-130a-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-130a
          Stem-loop Accession # MI0023317
          Stem-loop Sequence
          GUCUGUCCAGUGCCCCUUUUAUAUUGUACUACUGAUAACCCAGUUAUUAUAGCAGUGCAAUGUUAAAAGGGCAUUGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026210
          miRBase ID: ccr-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-130a
          Stem-loop Accession # MI0038974
          Stem-loop Sequence
          GCUCUUUUCACAUUGUGCUACUGUCUGCACCUAUUACUAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047628
          miRBase ID: dno-miR-130a-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-130a
          Stem-loop Accession # MI0008029
          Stem-loop Sequence
          GCUCUUUUCACAUUGUGCUACUGUCUGCACCUACCACUAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006631
          miRBase ID: cfa-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-130a
          Stem-loop Accession # MI0038633
          Stem-loop Sequence
          GCUCUUUUCACAUUGUGCUACUGUCUGCACUCGUCACAAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046981
          miRBase ID: cpo-miR-130a-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-130a
          Stem-loop Accession # MI0012790
          Stem-loop Sequence
          GAGCUCUUUUCACAUUGUGCUACUGUCUGCACCUACCACUAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013047
          miRBase ID: eca-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-130c-1
          Stem-loop Accession # MI0023774
          Stem-loop Sequence
          GCCCUUUUUAUGUUGUACUACUAGUGAUCCUGCACAAGAAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028515
          miRBase ID: mdo-miR-130c-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Stem-loop ID mdo-mir-130c-2
          Stem-loop Accession # MI0023803
          Stem-loop Sequence
          GCCCUUUUCUUGUUGCCCUGCUGAGCCAGCGAGGGGGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028515
          miRBase ID: mdo-miR-130c-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-130b
          Stem-loop Accession # MI0031336
          Stem-loop Sequence
          UGUCCGUCCACUGCCCUUUUUAUGUUGUACUACUAGUGAACAUGCACAAAAAGCAGUGCAAUGUUAAAAGGGCAUUGGCUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036690
          miRBase ID: oha-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-130b
          Stem-loop Accession # MI0006882
          Stem-loop Sequence
          UAGUACGGUUGAACAUUUCAUGAGUGGUAUCUGUCUAGUGCCCUUUUUAUGUUGUACUACUAGUGAUCUUGCACAAAAAGCAGUGCAAUGUUAAAAGGGCAUUGGCCAGUGAUACUGUUCACCUUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007159
          miRBase ID: oan-miR-130b-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-130a
          Stem-loop Accession # MI0040449
          Stem-loop Sequence
          GCUCUUUUCACAUUGUGCUACUGUCUGCACCUGUCACUAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049666
          miRBase ID: oga-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-130a
          Stem-loop Accession # MI0008527
          Stem-loop Sequence
          GCUGCUGGCCAGAGCUCUUUUCACAUUGUGCUACUGUCUGCACCUGUCACUAGCAGUGCAAUGUUAAAAGGGCAUUGGCCGUGUAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008025
          miRBase ID: ptr-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-130a
          Stem-loop Accession # MI0008217
          Stem-loop Sequence
          CGGGCCGGAGCUCUUUUCACAUUGUGCUACUGUCUGCACCUAUCACUAGCAGUGCAAUGUUAAAAGGGCAUUGGCCACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007758
          miRBase ID: ssc-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-130a
          Stem-loop Accession # MI0014822
          Stem-loop Sequence
          UGCUGCUGGCCAGAGCUCUUUUCACAUUGUGCUACUGUCUGCACCUGUCACUAGCAGUGCAAUGUUAAAAGGGCAUUGGCCGUGUAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015753
          miRBase ID: ppy-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-130
          Stem-loop Accession # MI0032388
          Stem-loop Sequence
          GCUCUUUUCACAUUGUGCUACUGUCUGCACCUACCACUAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039896
          miRBase ID: pal-miR-130-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-130d
          Stem-loop Accession # MI0030226
          Stem-loop Sequence
          AGAGGUGUCUGUCCACUGCCCUUUUUAUGUUGUACUACUAGUGAACACACACAAAAAGCAGUGCAAUGUUAAAAGGGCAUUGGCCAGCAAUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038932
          miRBase ID: pbv-miR-130d-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-130c
          Stem-loop Accession # MI0013808
          Stem-loop Sequence
          UCCUGCACCUCCUCUGUCCCUGAGUGGUGUCUGUCCAGUGCCCUUUUUAUGUUGUACUACUGGUGAUGAUGCACAAAAAGCAGUGCAAUGUUAAAAGGGCAUUGGCCAGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014581
          miRBase ID: tgu-miR-130c-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-130a
          Stem-loop Accession # MI0031241
          Stem-loop Sequence
          GCUCUUUUCACAUUGUGCUACUGUCUGCAUCUGUCACUAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036577
          miRBase ID: tch-miR-130a-3p
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-130a
          Stem-loop Accession # MI0004831
          Stem-loop Sequence
          CUGUCCAGUGCCCUUUUUAUGUUGUACUACUAGUGGUCACACACAAAAAAAGCAGUGCAAUGUUAAAAGGGCAUUGGCCAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003591
          miRBase ID: xtr-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-130a
          Stem-loop Accession # MI0001984
          Stem-loop Sequence
          UGUCUGUCCAGUGCCCCUUUUAUAUUGUACUACUGAUAACCCAGUUAUUAAAGCAGUGCAAUGUUAAAAGGGCAUUGGCCAGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001826
          miRBase ID: dre-miR-130a
          Mature miRNA Sequence: CAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306263_pri
          TaqMan™ Pri-miRNA Assay : Hs03303108_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10506
          Ambion® Pre-miR™ miRNA Precursor : PM10506
          mirVana® miRNA inhibitor : MH10506
          mirVana® miRNA mimic : MC10506
          TaqMan™ Advanced miRNA Assay : 477851_mir
          TaqMan™ Advanced miRNA Assay : mmu483331_mir
          TaqMan™ Advanced miRNA Assay : rno477851_mir
          FGMIR : TaqManTM Rodent MicroRNA Array A
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : TaqManTM Human MicroRNA Array A
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel


          miRBase Alias:

          Human: hsa-miR-130a (v17)
          Mouse: mmu-miR-130a (v17)
          Rat: rno-miR-130a (v18)
          Gallus gallus: gga-miR-130c (v18)
          Ochrobactrum anthropi: oan-miR-130b (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-130c (v19)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline