Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC19316
          Assay Name: mmu-miR-146a-3p
          miRBase Accession Number: MI0000170
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 43374397 - 43374461 [-] on Build GRCm38
          Mature miRNA Sequence: CCUGUGAAAUUCAGUUCUUCAG
          Species: Mouse, Pteropus alecto, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC19316

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000103, mir-146

          Mature miRNA Sequence:

          CCUGUGAAAUUCAGUUCUUCAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-146a-3p MIMAT0016989
          Pteropus alecto pal-miR-146a-3p MIMAT0040182
          Rhesus monkey mml-miR-146a-3p MIMAT0026824

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-146a
          Stem-loop Accession # MI0000170
          Stem-loop Sequence
          AGCUCUGAGAACUGAAUUCCAUGGGUUAUAUCAAUGUCAGACCUGUGAAAUUCAGUUCUUCAGCU
          Chromosome Location Chr. 11 - 43374397 - 43374461 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0016989
          miRBase ID: mmu-miR-146a-3p
          Mature miRNA Sequence: CCUGUGAAAUUCAGUUCUUCAG
          Chromosome Location: Chr. 11 - 43374397 - 43374461 [-] on Build GRCm38

          Pteropus alecto

          Stem-loop ID pal-mir-146a
          Stem-loop Accession # MI0032537
          Stem-loop Sequence
          UGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAAUUCAGUUCUUCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040182
          miRBase ID: pal-miR-146a-3p
          Mature miRNA Sequence: CCUGUGAAAUUCAGUUCUUCAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-146a
          Stem-loop Accession # MI0007634
          Stem-loop Sequence
          CCUAUGUGUAUCCUCAGCUUUGAGAACUGAAUUCCAUGGGUUGUGUCAGUGUCAGACCUGUGAAAUUCAGUUCUUCAGCUGGGAUAUCUCUGUCGUCGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026824
          miRBase ID: mml-miR-146a-3p
          Mature miRNA Sequence: CCUGUGAAAUUCAGUUCUUCAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306349_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19316
          Ambion® Pre-miR™ miRNA Precursor : PM19316
          TaqMan™ MicroRNA Assay : 463191_mat
          mirVana® miRNA inhibitor : MH19316
          TaqMan™ Advanced miRNA Assay : mmu481546_mir


          miRBase Alias:

          Mouse: mmu-miR-146a* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-hgw72:80/100.66.79.134:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline