Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC32942
          Assay Name: mmu-miR-181a-2-3p
          miRBase Accession Number: MI0000223
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 38852735 - 38852810 [+] on Build GRCm38
          Mature miRNA Sequence: ACCACCGACCGUUGACUGUACC
          Species: Mouse, Eptesicus fuscus, Pteropus alecto, Rabbit
          Product Type: mirVana® miRNA mimic
          Assay ID MC32942

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          ACCACCGACCGUUGACUGUACC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-181a-2-3p MIMAT0005443
          Eptesicus fuscus efu-miR-181f MIMAT0035008
          Pteropus alecto pal-miR-181a-2-3p MIMAT0039816
          Rabbit ocu-miR-181a-2-3p MIMAT0048277

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-181a-2
          Stem-loop Accession # MI0000223
          Stem-loop Sequence
          CCAUGGAACAUUCAACGCUGUCGGUGAGUUUGGGAUUCAAAAACAAAAAAACCACCGACCGUUGACUGUACCUUGG
          Chromosome Location Chr. 2 - 38852735 - 38852810 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0005443
          miRBase ID: mmu-miR-181a-2-3p
          Mature miRNA Sequence: ACCACCGACCGUUGACUGUACC
          Chromosome Location: Chr. 2 - 38852735 - 38852810 [+] on Build GRCm38

          Eptesicus fuscus

          Stem-loop ID efu-mir-181f
          Stem-loop Accession # MI0028693
          Stem-loop Sequence
          GACUCCAAGGAACAUUCAACGCUGUCGGUGAGUUUGGGAUUUGAAAAAACCACCGACCGUUGACUGUACCUUGGGCUCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035008
          miRBase ID: efu-miR-181f
          Mature miRNA Sequence: ACCACCGACCGUUGACUGUACC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-181a-2
          Stem-loop Accession # MI0032343
          Stem-loop Sequence
          AACAUUCAACGCUGUCGGUGAGUUUGGGAUUUGAAAAAACCACCGACCGUUGACUGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039816
          miRBase ID: pal-miR-181a-2-3p
          Mature miRNA Sequence: ACCACCGACCGUUGACUGUACC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-181a-2
          Stem-loop Accession # MI0039316
          Stem-loop Sequence
          AACAUUCAACGCUGUCGGUGAGUUUGGGAUUUGAAAAAACCACCGACCGUUGACUGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048277
          miRBase ID: ocu-miR-181a-2-3p
          Mature miRNA Sequence: ACCACCGACCGUUGACUGUACC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306417_pri
          TaqMan™ MicroRNA Assay : 473858_mat
          mirVana® miRNA inhibitor : MH32942

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0