Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH19432
          Assay Name: hsa-miR-130a-5p
          miRBase Accession Number: MI0000448
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 57641198 - 57641286 [+] on Build GRCh38
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Species: Human, Mouse, Rat, Chrysemys picta, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Monodelphis domestica, Ophiophagus hannah, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH19432

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000034, mir-130

          Mature miRNA Sequence:

          GCUCUUUUCACAUUGUGCUACU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-130a-5p MIMAT0004593
          Mouse mmu-miR-130a-5p MIMAT0016983
          Rat rno-miR-130a-5p MIMAT0017121
          Chrysemys picta cpi-miR-130a-5p MIMAT0037741
          Cricetulus griseus cgr-miR-130a-5p MIMAT0023754
          Dasypus novemcinctus dno-miR-130a-5p MIMAT0047627
          View More Rows
          Goat chi-miR-130a-5p MIMAT0035943
          Guinea pig cpo-miR-130a-5p MIMAT0046980
          Monodelphis domestica mdo-miR-130a-5p MIMAT0026664
          Ophiophagus hannah oha-miR-130c-5p MIMAT0036691
          Rabbit ocu-miR-130a-5p MIMAT0048217
          Rhesus monkey mml-miR-130a-5p MIMAT0026566

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-130a
          Stem-loop Accession # MI0000448
          Stem-loop Sequence
          UGCUGCUGGCCAGAGCUCUUUUCACAUUGUGCUACUGUCUGCACCUGUCACUAGCAGUGCAAUGUUAAAAGGGCAUUGGCCGUGUAGUG
          Chromosome Location Chr. 11 - 57641198 - 57641286 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004593
          miRBase ID: hsa-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: Chr. 11 - 57641198 - 57641286 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-130a
          Stem-loop Accession # MI0000156
          Stem-loop Sequence
          GAGCUCUUUUCACAUUGUGCUACUGUCUAACGUGUACCGAGCAGUGCAAUGUUAAAAGGGCAUC
          Chromosome Location Chr. 2 - 84741115 - 84741178 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0016983
          miRBase ID: mmu-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: Chr. 2 - 84741115 - 84741178 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-130a
          Stem-loop Accession # MI0000903
          Stem-loop Sequence
          UGCUGCUGGCCGGAGCUCUUUUCACAUUGUGCUACUGUCUACACGUGUACCGAGCAGUGCAAUGUUAAAAGGGCAUCGGCCUUGUAGU
          Chromosome Location Chr. 3 - 72141037 - 72141124 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017121
          miRBase ID: rno-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: Chr. 3 - 72141037 - 72141124 [-] on Build Rnor_6.0

          Chrysemys picta

          Stem-loop ID cpi-mir-130a
          Stem-loop Accession # MI0029467
          Stem-loop Sequence
          CACGCCGCCGUCCUGGGCUCUUUUCACAUUGUGCUACUGUCUGAGCCCCUGCCAAGCAGUGCAAUGUCAAAAGGGCAUCGGGCAGGCGGUGCCGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037741
          miRBase ID: cpi-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-130a
          Stem-loop Accession # MI0020394
          Stem-loop Sequence
          GCUGGCCGGAGCUCUUUUCACAUUGUGCUACUGUCUUCACCUAUAACGAGCAGUGCAAUGUUAAAAGGGCAUCGGCCUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023754
          miRBase ID: cgr-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-130a
          Stem-loop Accession # MI0038974
          Stem-loop Sequence
          GCUCUUUUCACAUUGUGCUACUGUCUGCACCUAUUACUAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047627
          miRBase ID: dno-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-130a
          Stem-loop Accession # MI0030616
          Stem-loop Sequence
          UGCUGCGGGCCGGAGCUCUUUUCACAUUGUGCUACUGUCUGCGCCUGUCACUAGCAGUGCAAUGUUAAAAGGGCAUUGGCCGCGCAGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035943
          miRBase ID: chi-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-130a
          Stem-loop Accession # MI0038633
          Stem-loop Sequence
          GCUCUUUUCACAUUGUGCUACUGUCUGCACUCGUCACAAGCAGUGCAAUGUUAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046980
          miRBase ID: cpo-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-130a
          Stem-loop Accession # MI0005295
          Stem-loop Sequence
          UGGCCAGAGCUCUUUUCACAUUGUGCUACUGUCUGCACCUAUCACUAGCAGUGCAAUGUAAAAAGGGCAUUGGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026664
          miRBase ID: mdo-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-130c
          Stem-loop Accession # MI0031337
          Stem-loop Sequence
          UUGGGCACGGCCCCUGCCCGAGGCUCUUUUCACAUUGUGCUACUGAGCCCAACCAAGCAGUGCAAUGUAAAAAGGGCAUUGGGUUGGUGGCCACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036691
          miRBase ID: oha-miR-130c-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-130a
          Stem-loop Accession # MI0039285
          Stem-loop Sequence
          GCUCUUUUCACAUUGUGCUACUGUCUGCGCCUAUCACGAGCAGUGCAAUGUCAAAAGGGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048217
          miRBase ID: ocu-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-130a
          Stem-loop Accession # MI0002524
          Stem-loop Sequence
          UGCUGCUGGCCAGAGCUCUUUUCACAUUGUGCUACUGUCUGCACCUGUCACUAGCAGUGCAAUGUUAAAAGGGCAUUGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026566
          miRBase ID: mml-miR-130a-5p
          Mature miRNA Sequence: GCUCUUUUCACAUUGUGCUACU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303108_pri
          TaqMan™ Pri-miRNA Assay : Mm03306263_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19432
          Ambion® Pre-miR™ miRNA Precursor : PM19432
          TaqMan™ MicroRNA Assay : 462691_mat
          mirVana® miRNA mimic : MC19432
          TaqMan™ Advanced miRNA Assay : rno481319_mir
          TaqMan™ Advanced miRNA Assay : 483130_mir
          TaqMan™ Advanced miRNA Assay : mmu482684_mir


          miRBase Alias:

          Mouse: mmu-miR-130a* (v17)
          Rat: rno-miR-130a* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline