Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03306867_pri
          Assay Name:
          mmu-mir-23a
          Stem-loop ID:
          mmu-mir-23a mmu-mir-27a
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.8: 84208518 - 84208592 [+] on Build GRCm38
          Stem-loop Location:
          Chr.8: 84208518 - 84208592 [+] on Build GRCm38 Chr.8: 84208672 - 84208758 [+] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-23a-3p mmu-miR-23a-5p mmu-miR-27a-3p mmu-miR-27a-5p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03306867_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          88

          Chromosome Location:

          Chr. 8 - 84208375 - 84208375 [-] on 84208375 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-23a-3p MIMAT0000532
          Mouse mmu-miR-23a-5p MIMAT0017019
          Mouse mmu-miR-27a-3p MIMAT0000537
          Mouse mmu-miR-27a-5p MIMAT0004633

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-23a
          Stem-loop Accession # MI0000571
          Stem-loop Sequence
          CGGACGGCUGGGGUUCCUGGGGAUGGGAUUUGAUGCCAGUCACAAAUCACAUUGCCAGGGAUUUCCAACUGACCC
          Chromosome Location Chr. 8 - 84208518 - 84208592 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000532
          miRBase ID: mmu-miR-23a-3p
          Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC
          Chromosome Location: Chr. 8 - 84208518 - 84208592 [+] on Build GRCm38

          miRBase Accession #: MIMAT0017019
          miRBase ID: mmu-miR-23a-5p
          Mature miRNA Sequence: GGGGUUCCUGGGGAUGGGAUUU
          Chromosome Location: Chr. 8 - 84208518 - 84208592 [+] on Build GRCm38

          Gene Family ID MIPF0000027, mir-23
          Distance to Amplicon 115 bases
          Stem-loop ID mmu-mir-27a
          Stem-loop Accession # MI0000578
          Stem-loop Sequence
          UGGCCUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAAGGUCCACAGCAAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCUGGACCC
          Chromosome Location Chr. 8 - 84208672 - 84208758 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000537
          miRBase ID: mmu-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: Chr. 8 - 84208672 - 84208758 [+] on Build GRCm38

          miRBase Accession #: MIMAT0004633
          miRBase ID: mmu-miR-27a-5p
          Mature miRNA Sequence: AGGGCUUAGCUGCUUGUGAGCA
          Chromosome Location: Chr. 8 - 84208672 - 84208758 [+] on Build GRCm38

          Gene Family ID MIPF0000036, mir-27
          Distance to Amplicon 269 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10939
          Ambion® Pre-miR™ miRNA Precursor : PM10939
          TaqMan™ MicroRNA Assay : 000408
          mirVana® miRNA inhibitor : MH10939
          mirVana® miRNA mimic : MC10939
          TaqMan™ Advanced miRNA Assay : 478384_mir
          TaqMan™ Advanced miRNA Assay : mmu478384_mir
          TaqMan™ Advanced miRNA Assay : rno478384_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM13096
          Ambion® Pre-miR™ miRNA Precursor : PM13096
          TaqMan™ MicroRNA Assay : 002445
          mirVana® miRNA inhibitor : MH13096
          mirVana® miRNA mimic : MC13096
          TaqMan™ Advanced miRNA Assay : rno481015_mir
          TaqMan™ Advanced miRNA Assay : 477998_mir
          TaqMan™ Advanced miRNA Assay : mmu481015_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10644
          Ambion® Pre-miR™ miRNA Precursor : PM10644
          TaqMan™ MicroRNA Assay : 000399
          mirVana® miRNA inhibitor : MH10644
          mirVana® miRNA mimic : MC10644
          TaqMan™ Advanced miRNA Assay : 478532_mir
          TaqMan™ Advanced miRNA Assay : mmu478532_mir
          TaqMan™ Advanced miRNA Assay : rno478532_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12940
          Ambion® Pre-miR™ miRNA Precursor : PM12940
          TaqMan™ MicroRNA Assay : 002439
          mirVana® miRNA inhibitor : MH12940
          mirVana® miRNA mimic : MC12940
          TaqMan™ Advanced miRNA Assay : 478782_mir
          TaqMan™ Advanced miRNA Assay : mmu482713_mir
          TaqMan™ Advanced miRNA Assay : rno481350_mir


          miRBase Alias:

          Mouse: mmu-mir-27a (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline