Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10939
          Assay Name: hsa-miR-27a-3p
          miRBase Accession Number: MI0000085
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 13836440 - 13836517 [-] on Build GRCh38
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Astatotilapia burtoni, Callithrix jacchus, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Horse, Metriaclima zebra, Monodelphis domestica, Neolamprologus brichardi, Oreochromis niloticus, Ornithorhynchus anatinus, Pig, Pundamilia nyererei, Python bivittatus, Saimiri boliviensis, Sheep, Xenopus tropicalis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10939

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000036, mir-27

          Mature miRNA Sequence:

          UUCACAGUGGCUAAGUUCCGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-27a-3p MIMAT0000084
          Mouse mmu-miR-27a-3p MIMAT0000537
          Rat rno-miR-27a-3p MIMAT0000799
          Alligator mississippiensis ami-miR-27a-3p MIMAT0038068
          Anolis carolinensis aca-miR-27a-3p MIMAT0021906
          Astatotilapia burtoni abu-miR-27a MIMAT0042109
          View More Rows
          Callithrix jacchus cja-miR-27a MIMAT0039518
          Chrysemys picta cpi-miR-27a-3p MIMAT0037665
          Cricetulus griseus cgr-miR-27a-3p MIMAT0023875
          Cyprinus carpio ccr-miR-27a MIMAT0026282
          Dasypus novemcinctus dno-miR-27a-3p MIMAT0047544
          Horse eca-miR-27a MIMAT0012988
          Metriaclima zebra mze-miR-27a MIMAT0042337
          Monodelphis domestica mdo-miR-27a-3p MIMAT0004174
          Neolamprologus brichardi nbr-miR-27a MIMAT0042530
          Oreochromis niloticus oni-miR-27a MIMAT0042710
          Ornithorhynchus anatinus oan-miR-27a-3p MIMAT0007063
          Pig ssc-miR-27a MIMAT0002148
          Pundamilia nyererei pny-miR-27a MIMAT0042914
          Python bivittatus pbv-miR-27a-3p MIMAT0038862
          Saimiri boliviensis sbo-miR-27a MIMAT0049478
          Sheep oar-miR-27a MIMAT0030053
          Xenopus tropicalis xtr-miR-27a MIMAT0003570

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-27a
          Stem-loop Accession # MI0000085
          Stem-loop Sequence
          CUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAGGGUCCACACCAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCCAG
          Chromosome Location Chr. 19 - 13836440 - 13836517 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000084
          miRBase ID: hsa-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: Chr. 19 - 13836440 - 13836517 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-27a
          Stem-loop Accession # MI0000578
          Stem-loop Sequence
          UGGCCUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAAGGUCCACAGCAAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCUGGACCC
          Chromosome Location Chr. 8 - 84208672 - 84208758 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000537
          miRBase ID: mmu-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: Chr. 8 - 84208672 - 84208758 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-27a
          Stem-loop Accession # MI0000860
          Stem-loop Sequence
          UGGCCUGUGGAGCAGGGCUUAGCUGCUUGUGAGCAAGGUCUACAGCAAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCUGGACCC
          Chromosome Location Chr. 19 - 25318736 - 25318822 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000799
          miRBase ID: rno-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: Chr. 19 - 25318736 - 25318822 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-27a
          Stem-loop Accession # MI0029684
          Stem-loop Sequence
          GAAGCCAGGGCACAGGGCUUAGCUCACCUGUGAACAGAGUUAGCGUUGCAUCAUGUUCACAGUGGCUAAGUUCCGCCUCCUGGUGUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038068
          miRBase ID: ami-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-27a
          Stem-loop Accession # MI0018837
          Stem-loop Sequence
          AGCCGUAGGGCUUAGCCCACCUGUGAACAGCAUUGGAUUCAGCCAUGUUCACAGUGGCUAAGUUCCGCUGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021906
          miRBase ID: aca-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-27a
          Stem-loop Accession # MI0033812
          Stem-loop Sequence
          AGGACUUAGCUCGCUCCGUGAACAGUUCAGUGAAGUCCUGUGUUCACAGUGGCUAAGUUCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042109
          miRBase ID: abu-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-27a
          Stem-loop Accession # MI0032046
          Stem-loop Sequence
          GAUGCCGCCUGGGGACGGGGUGGCAGAGAGCCCCGACGCCUGUGCCUGGCCUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAGGGUUCACUCCAAGUCAUGUUCACAGUGGCUAAGUUCCGCCCCCCAGGCCCUCACCUCCUCCGGCCCUGCCGCCUGUCCCCCAAUGCUGCCUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039518
          miRBase ID: cja-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-27a
          Stem-loop Accession # MI0029418
          Stem-loop Sequence
          GAUACCGGGGUGCAGGGCUUAGCUCACCUGUGAACAGAGUUCGCAUCCGUAUCGUGUUCACAGUGGCUAAGUUCCGCCCCCCGGGGUCCGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037665
          miRBase ID: cpi-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-27a
          Stem-loop Accession # MI0020473
          Stem-loop Sequence
          CCUGAGGAGCAGGGCUUAGCUGCUUGUGAGCAAGGUCCGCAGCAAGUCGUGUUCACAGUGGCUAAGUUCCGCUCCCCCGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023875
          miRBase ID: cgr-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-27a
          Stem-loop Accession # MI0023383
          Stem-loop Sequence
          GAGGUGCAGGACUUAGCUCUCUCUGUGAACAGAUCUCGGAUAUCCUAUGUUCACAGUGGCUAAGUUCCGCUCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026282
          miRBase ID: ccr-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-27a
          Stem-loop Accession # MI0038927
          Stem-loop Sequence
          AGGGCUUAGCUGCUUGUGAGCAGGGUCUGCAGCAAGUCGUGUUCACAGUGGCUAAGUUCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047544
          miRBase ID: dno-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-27a
          Stem-loop Accession # MI0012739
          Stem-loop Sequence
          CUGGGGAGCAGGGCUUAGCUGCUUGUGAGCAGGGUCCACACCGAGUCAUGUUCACAGUGGCUAAGUUCCGCCCCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012988
          miRBase ID: eca-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-27a
          Stem-loop Accession # MI0034108
          Stem-loop Sequence
          AGGACUUAGCUCGCUCCGUGAACAGUUCAGUGAAGUCCUGUGUUCACAGUGGCUAAGUUCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042337
          miRBase ID: mze-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-27a
          Stem-loop Accession # MI0005364
          Stem-loop Sequence
          UGGCUUGAGGAGCAGGACUUAGCUGCCUUGUGAACAGAGUCAGCAUCAUAUUGUGUUCACAGUGGCUAAGUUCCGCUCCCCUUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004174
          miRBase ID: mdo-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-27a
          Stem-loop Accession # MI0034375
          Stem-loop Sequence
          AGGACUUAGCUCGCUCCGUGAACAGUUCAGUGAAGUCCUGUGUUCACAGUGGCUAAGUUCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042530
          miRBase ID: nbr-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-27a
          Stem-loop Accession # MI0034624
          Stem-loop Sequence
          AGGACUUAGCUCGCUCCGUGAACAGUUCAGUGAAAUCCUGUGUUCACAGUGGCUAAGUUCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042710
          miRBase ID: oni-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-27a
          Stem-loop Accession # MI0006825
          Stem-loop Sequence
          GGUCCGGCGGAGCAGGGCUUAGCUGGCCUGUGAACAGUUAGUUCCCUGCGUGUUCACAGUGGCUAAGUUCCGCCCCUCCAGGGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007063
          miRBase ID: oan-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-27a
          Stem-loop Accession # MI0002442
          Stem-loop Sequence
          UGGCCUGGGGAGCAGGGCUUAGCUGCUUGUGAGCAGGUCCACAGCAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCCUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002148
          miRBase ID: ssc-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-27a
          Stem-loop Accession # MI0034890
          Stem-loop Sequence
          AGGACUUAGCUCGCUCCGUGAACAGUUCAGUGAAGUCCUGUGUUCACAGUGGCUAAGUUCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042914
          miRBase ID: pny-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-27a
          Stem-loop Accession # MI0030177
          Stem-loop Sequence
          GACUGCCUAGGGUAGGGCUUAGCUCACUUGUGAACAGCGUUUGGUUCAGCCGUGUUCACAGUGGCUAAGUUCCGCCACUUGGAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038862
          miRBase ID: pbv-miR-27a-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Saimiri boliviensis

          Stem-loop ID sbo-mir-27a
          Stem-loop Accession # MI0040235
          Stem-loop Sequence
          AGGGCUUAGCUGCUUGUGAGCAGGGUUCACUCCAAGUCAUGUUCACAGUGGCUAAGUUCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049478
          miRBase ID: sbo-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-27a
          Stem-loop Accession # MI0025275
          Stem-loop Sequence
          UGGCCUGGGGAGCAGGGCUUAGCUGCUUGUGAGCAGGUCCACAUCAAGUCGUGUUCACAGUGGCUAAGUUCCGCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030053
          miRBase ID: oar-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-27a
          Stem-loop Accession # MI0004809
          Stem-loop Sequence
          CGGCGGCCCAGGGCUUAGCUGUAUUGUGAGCACUGUCACUCUCGCACCUGUUCACAGUGGCUAAGUUCCGCGCCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003570
          miRBase ID: xtr-miR-27a
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCCGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04270764_pri
          TaqMan™ Pri-miRNA Assay : Rn03465981_pri
          TaqMan™ Pri-miRNA Assay : Mm03306867_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10939
          Ambion® Pre-miR™ miRNA Precursor : PM10939
          TaqMan™ MicroRNA Assay : 000408
          mirVana® miRNA mimic : MC10939
          TaqMan™ Advanced miRNA Assay : 478384_mir
          TaqMan™ Advanced miRNA Assay : mmu478384_mir
          TaqMan™ Advanced miRNA Assay : rno478384_mir


          miRBase Alias:

          Human: hsa-miR-27a (v17)
          Mouse: mmu-miR-27a (v17)
          Rat: rno-miR-27a (v18)
          Aeromonas caviae Ae398: aca-miR-27a (v18)
          Monodelphis domestica: mdo-miR-27a (v19)
          Ochrobactrum anthropi: oan-miR-27a (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline