Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481431_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-9b-5p
          Stem-loop Accession Number: MI0015430
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 187582790 - 187582897 [-] on Build Rnor_6.0
          Mature miRNA Sequence: UUCGGUUAUCUAGCUUUAU
          Species: Rat
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481431_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000014, mir-9

          Mature miRNA Sequence:

          UUCGGUUAUCUAGCUUUAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-9b-5p MIMAT0017835

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-9b-1
          Stem-loop Accession # MI0015430
          Stem-loop Sequence
          GCGCAGUGUAUGGGGUUAUUUUUACUUUCGGUUAUCUAGCUUUAUGAAGACUCCACACCACUCAUACAGCUAGAUAACCAAAGAUAACAACCAACCCCGCCUCCUGGC
          Chromosome Location Chr. 2 - 187582790 - 187582897 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017835
          miRBase ID: rno-miR-9b-5p
          Mature miRNA Sequence: UUCGGUUAUCUAGCUUUAU
          Chromosome Location: Chr. 2 - 11884658 - 11884775 [-] on Build Rnor_6.0

          Stem-loop ID rno-mir-9b-2
          Stem-loop Accession # MI0015439
          Stem-loop Sequence
          AGCACUCCUCGGUGAUCUUGAAGGAGUUUUUACUUUCGGUUAUCUAGCUUUAUGAAGACCAAUACACUCAUACAGCUAGAUAACCAAAGAUAACAACUCGCUUCCCUCACAAGGCUGU
          Chromosome Location Chr. 2 - 11884658 - 11884775 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017835
          miRBase ID: rno-miR-9b-5p
          Mature miRNA Sequence: UUCGGUUAUCUAGCUUUAU
          Chromosome Location: Chr. 2 - 11884658 - 11884775 [-] on Build Rnor_6.0

          Stem-loop ID rno-mir-9b-3
          Stem-loop Accession # MI0015426
          Stem-loop Sequence
          GCACGCAGAAGUUGUUAGAGUCAUUUCUACUUUCGGUUAUCUAGCUUUAUGACGGCUCUGUGGCACUCAUACAGCUAGAUAACCAAAGAGAGAAACGGGCCUCCCAUUCGCCGGC
          Chromosome Location Chr. 1 - 141229342 - 141229456 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017835
          miRBase ID: rno-miR-9b-5p
          Mature miRNA Sequence: UUCGGUUAUCUAGCUUUAU
          Chromosome Location: Chr. 2 - 11884658 - 11884775 [-] on Build Rnor_6.0


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465966_pri
          TaqMan™ Pri-miRNA Assay : Rn03464510_pri
          TaqMan™ Pri-miRNA Assay : Rn03465960_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM20226
          Ambion® Pre-miR™ miRNA Precursor : PM20226
          TaqMan™ MicroRNA Assay : 462133_mat
          mirVana® miRNA inhibitor : MH20226
          mirVana® miRNA mimic : MC20226

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline