Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • CARD11 Antibodies

          Proteintech

          CARD11 Polyclonal Antibody

          4 References
          View all (17) CARD11 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CARD11 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          CARD11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CARD11 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CARD11 Antibody (21741-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed L02 cells using CARD11 antibody (Product # 21741-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CARD11 Antibody in Immunocytochemistry (ICC/IF)
          CARD11 Antibody in Immunocytochemistry (ICC/IF)
          CARD11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CARD11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CARD11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CARD11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CARD11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CARD11 Antibody in Western Blot (WB)
          CARD11 Antibody in Western Blot (WB)
          CARD11 Antibody in Western Blot (WB)
          CARD11 Polyclonal Antibody

          Product Details

          21741-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CARD11 Fusion Protein Ag16468 (805-1154 aa encoded by BC111719)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.26 mg/mL

          Amount

          39 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: DTMYQDRHE WLCARVDPFT DHDLDMGTIP SYSRAQQLLL VKLQRLMHRG SREEVDGTHH TLRALRNTLQ PEEALSTSDP RVSPRLSRAS FLFGQLLQFV SRSENKYKRM NSNERVRIIS GSPLGSLARS SLDATKLLTE KQEELDPESE LGKNLSLIPY SLVRAFYCER RRPVLFTPTV LAKTLVQRLL NSGGAMEFTI CKSDIVTRDE FLRRQKTETI IYSREKNPNA FECIAPANIE AVAAKNKHCL LEAGIGCTRD LIKSNIYPIV LFIRVCEKNI KRFRKLLPRP ETEEEFLRVC RLKEKELEAL PCLYATVEPD MWGSVEELLR VVKDKIGEEQ RKTIWVDEDQ L (805-1154 aa encoded by BC111719)

          Target Information

          The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein has a domain structure similar to that of CARD14 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: bcl10-interacting maguk protein 3; CARD-containing MAGUK protein 1; CARD11; CARD/MAGUK family member; Carma 1; caspase recruitment domain family, member 11; Caspase recruitment domain-containing protein 11; MAGUK; MGC133069

          View more View less

          Gene Aliases: 0610008L17Rik; 2410011D02Rik; BENTA; BIMP3; CARD11; CARMA1; IMD11; IMD11A; PPBL

          View more View less

          UniProt ID: (Human) Q9BXL7, (Mouse) Q8CIS0

          View more View less

          Entrez Gene ID: (Human) 84433, (Mouse) 108723, (Rat) 100363332

          View more View less

          Function(s)
          guanylate kinase activity protein binding CARD domain binding
          Process(es)
          positive regulation of cytokine production signal transduction positive regulation of B cell proliferation T cell costimulation positive regulation of T cell proliferation regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling thymic T cell selection positive regulation of interleukin-2 biosynthetic process regulation of B cell differentiation regulation of T cell differentiation regulation of immune response positive regulation of T cell activation positive regulation of NF-kappaB transcription factor activity immune system process I-kappaB kinase/NF-kappaB signaling B cell differentiation positive regulation of interleukin-2 production CD4-positive, alpha-beta T cell proliferation TORC1 signaling B cell proliferation T cell activation GMP metabolic process lymphocyte activation GDP metabolic process homeostasis of number of cells positive regulation of T cell receptor signaling pathway protein homooligomerization positive regulation of CD4-positive, alpha-beta T cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline