Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Cytokeratin HMW Antibodies

          Invitrogen

          Cytokeratin HMW Monoclonal Antibody (34BetaE12)

          16 References
          View all (4) Cytokeratin HMW antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Cytokeratin HMW Monoclonal Antibody (34BetaE12)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          Cytokeratin HMW Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Cytokeratin HMW Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cytokeratin HMW Antibody (MA5-12135) in ICC/IF

          Immunofluorescence analysis of Cytokeratin HMW was done on 70% confluent log phase A431 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with of Cytokeratin HMW (34BetaE12) Mouse Monoclonal Antibody (Product # MA5-12135) at 1:250 dilution in0.1% BSA and inc... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cytokeratin HMW Antibody in Immunocytochemistry (ICC/IF)
          Cytokeratin HMW Antibody in Immunocytochemistry (ICC/IF)
          Cytokeratin HMW Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cytokeratin HMW Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cytokeratin HMW Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cytokeratin HMW Antibody in Western Blot (WB)
          Cytokeratin HMW Antibody in Western Blot (WB)
          Cytokeratin HMW Monoclonal Antibody (34BetaE12)

          Product Details

          MA5-12135

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:10,000
          -

          Immunohistochemistry (IHC)

          -
          View 13 publications 13 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:100
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:250
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Horse, Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          34BetaE12

          Immunogen

          Solubilized keratin extracted from human stratum corneum
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          tissue culture supernatant

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10983918

          Product Specific Information

          MA5-12135 targets Cytokeratin HMW and it recognizes keratin 1, 5, 10, and 14 in the Moll catalog (MW 68kDa, 58kDa, 56.5kDa, and 50kDa), respectively. The antibody has been tested in ICC/IF, WB and IHC (P) applications and shows reactivity with Equine, Human, Non-human primate, and Rabbit samples. This antibody does not react with rat tissue in Western blot applications.

          In normal cells, Cytokeratin HMW labels squamous, ductal and other complex epithelia. It reacts with benign small-acinir lesions of the prostate and does not react with hepatocytes, pancreatic acinar cells, proximal renal tubes or endometrial glands. In tumor cells, this antibody is reactive with both squamous and ductal neoplasms and variably with those derived from simple epithelium.

          The MA5-12135 immunogen is solubilized keratin extracted from human stratum corneum.

          Target Information

          Keratins are heteropolymeric structural proteins that form the intermediate filament. Cytokeratins are intermediate filament keratins found in the intracytoplasmic cytoskeleton of epithelial tissue. There are two types of cytokeratins - the low weight, acidic type I and the high weight, basic type II. Cytokeratins are usually found in pairs - one type I and one type II. KRT1 regulates the activity of kinases such as PKC and SRC via binding to integrin beta-1 (ITB1) and the receptor of activated protein C kinase I (RACK1). In complex with C1QBP it is a high affinity receptor for kininogen-1/HMWK. KRT1 is expressed in the spinous and granular layers of the epidermis with family member KRT10. Mutations affecting the gene result in bullous congenital ichthyosiform erythroderma. KRT5 is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations affecting the gene result in epidermolysis bullosa simplex diseases. KRT10 plays a role in the establishment of the epidermal barrier on plantar skin. It also acts as a mediator of S.aureus adherence to desquamated nasal epithelial cells via clfB - thus it plays a role in nasal colonization. Mutations affecting the gene result in epidermolytic hyperkeratosis. KRT14's nonhelical tail domain is involved in promoting KRT5-KRT14 filaments to self-organize into large bundles and enhances the mechanical properties involved in resilience of keratin intermediate filaments in vitro. Mutations affecting the gene result in epidermolysis bullosa simplex.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 58 kDa cytokeratin; 67 kDa cytokeratin; CK-1; CK-10; CK-14; CK-5; cytokeratin 1; cytokeratin 10; cytokeratin 14; Cytokeratin-1; Cytokeratin-10; Cytokeratin-14; Cytokeratin-5; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolytic hyperkeratosis 1; Hair alpha protein; K1; K10; K14; K5; keratin 1, type II; keratin 10, type I; keratin 14, type I; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin K5; keratin type II; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 14; Keratin, type II cytoskeletal 1; Keratin, type II cytoskeletal 5; Keratin-1; Keratin-10; Keratin-14; Keratin-5; KRT5; type II keratin subunit protein; Type-II keratin Kb1; Type-II keratin Kb5; unnamed protein product

          View more View less

          Gene Aliases: AEI2; BCIE; BIE; CK1; CK10; CK14; CK5; DDD; DDD1; EBS1; EBS1A; EBS1B; EBS1C; EBS1D; EBS2; EBS2A; EBS2B; EBS2C; EBS2D; EBS2E; EBS2F; EBS3; EBS4; EHK; EHK1; EHK2; EHK2A; EHK2B; EPPK; IHL; K1; K10; K14; K5; KPP; KRT1; KRT10; KRT14; KRT1A; KRT5; KRT5A; KRTA; NEPPK; NFJ

          View more View less

          UniProt ID: (Human) P04264, (Human) P13645, (Human) P02533, (Human) P13647

          View more View less

          Entrez Gene ID: (Human) 3848, (Human) 3858, (Human) 3861, (Human) 3852

          View more View less

          Function(s)
          protein binding carbohydrate binding structural constituent of epidermis signaling receptor activity protein heterodimerization activity structural molecule activity structural constituent of cytoskeleton keratin filament binding scaffold protein binding
          Process(es)
          morphogenesis of an epithelium epidermis development response to radiation keratinocyte differentiation hair cycle intermediate filament organization intermediate filament bundle assembly stem cell differentiation peptide cross-linking positive regulation of epidermis development protein heterotetramerization complement activation, lectin pathway response to oxidative stress keratinization fibrinolysis regulation of angiogenesis negative regulation of inflammatory response establishment of skin barrier response to mechanical stimulus regulation of cell migration regulation of protein localization intermediate filament polymerization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline