Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Endothelin B Receptor Antibodies

          Invitrogen

          Endothelin B Receptor Polyclonal Antibody

          3 Published Figures
          9 References
          View all (17) Endothelin B Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Endothelin B Receptor Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (3)
          Endothelin B Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Endothelin B Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Endothelin B Receptor Antibody (PA3-066) in ICC/IF

          Immnoctyochemistry of ETB in ETB-transfected cells {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Endothelin B Receptor Antibody in Immunocytochemistry (ICC/IF)
          Endothelin B Receptor Antibody in Immunohistochemistry (IHC)
          Endothelin B Receptor Antibody in Western Blot (WB)
          Endothelin B Receptor Antibody in Multiplex Immunohistochemistry (mIHC)
          Endothelin B Receptor Antibody in Western Blot (WB)
          Endothelin B Receptor Antibody in Western Blot (WB)
          Endothelin B Receptor Antibody in Western Blot (WB)
          Endothelin B Receptor Polyclonal Antibody

          Product Details

          PA3-066

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000
          View 6 publications 6 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Multiplex Immunohistochemistry (mIHC)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:2,000
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:5,000
          View 1 publication 1 publication

          Immunoelectrophoresis (IE)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          COOH-terminal amino acids of mouse ETB (424-442) KANDHGYDNFRSSNKYSSS coupled to KLH
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2540493

          Product Specific Information

          PA3-066 detects Endothelin Receptor B from human, mouse, and rat samples.

          PA3-066 has been successfully used in Western blot, immunocytochemistry, and immunohistochemistry (paraffin) procedures. By Western blot, this antibody detects a 47 kDa protein corresponding to mouse Endothelin Receptor B.

          The PA3-066 antigen is the COOH-terminal amino acids of mouse ETB (424-442) KANDHGYDNFRSSNKYSSS coupled to KLH.

          Target Information

          Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: AU022549; endothelin B receptor; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; endothelin-B receptor; ET-B; Et-b receptor; Et-b-r; ET-BR; ET-RB; Etb receptor; Etb-svr; G-protein coupled receptor; Hirschsprung disease 2; piebald; RP11-318G21.1; unnamed protein product

          View more View less

          Gene Aliases: ABCDS; Ednra; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETR-b; ETRB; HSCR; HSCR2; Sox10m1; WS4A

          View more View less

          UniProt ID: (Human) P24530, (Rat) P21451, (Mouse) P48302

          View more View less

          Entrez Gene ID: (Human) 1910, (Rat) 50672, (Mouse) 13618

          View more View less

          Function(s)
          G-protein coupled receptor activity endothelin receptor activity protein binding peptide hormone binding type 1 angiotensin receptor binding receptor activity signal transducer activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter neural crest cell migration renin secretion into blood stream regulation of heart rate regulation of pH signal transduction cell surface receptor signaling pathway G-protein coupled receptor signaling pathway negative regulation of adenylate cyclase activity phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration nervous system development peripheral nervous system development posterior midgut development regulation of blood pressure positive regulation of cell proliferation gene expression negative regulation of neuron maturation vein smooth muscle contraction vascular smooth muscle contraction calcium-mediated signaling cGMP-mediated signaling heparin metabolic process melanocyte differentiation regulation of fever generation aldosterone metabolic process response to lipopolysaccharide enteric smooth muscle cell differentiation positive regulation of urine volume renal sodium excretion epithelial fluid transport vasoconstriction vasodilation negative regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling pigmentation developmental pigmentation macrophage chemotaxis response to pain enteric nervous system development regulation of epithelial cell proliferation negative regulation of protein metabolic process sodium ion homeostasis canonical Wnt signaling pathway positive regulation of penile erection establishment of endothelial barrier renal sodium ion absorption calcium ion transmembrane transport cellular response to lipopolysaccharide protein transmembrane transport glomerular visceral epithelial cell differentiation endothelin receptor signaling pathway renal albumin absorption neuroblast migration chordate pharynx development response to sodium phosphate response to endothelin positive regulation of protein phosphorylation aging response to organic cyclic compound sensory perception of pain negative regulation of cellular protein metabolic process positive regulation of renal sodium excretion regulation of sensory perception of pain
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline