Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PMCA ATPase Antibodies

          Invitrogen

          PMCA ATPase Polyclonal Antibody

          1 Reference
          View all (4) PMCA ATPase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PMCA ATPase Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          PMCA ATPase Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PMCA ATPase Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PMCA ATPase Antibody (PA5-107073) in WB

          Western Blot analysis of PMCA ATPase in human 721_B cells using a PMCA ATPase Polyclonal Antibody (Product # PA5-107073) diluted to 0.2-1 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PMCA ATPase Antibody in Western Blot (WB)
          PMCA ATPase Polyclonal Antibody

          Product Details

          PA5-107073

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.2-1 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide directed towards the middle region of human ATP2B4 (aa 936-985)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.4, with 2% sucrose

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2817789

          Product Specific Information

          Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%

          Immunogen sequence: FAGEKFFDID SGRKAPLHSP PSQHYTIVFN TFVLMQLFNE INSRKIHGEK

          Target Information

          The calcium pump of the plasma membrane (PMCA ATPase) pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the sarcoplasmic and endoplasmic reticulum calcium (SERCA) ATPase and the sodium/potassium pump. Studies indicate that there are four known isoforms of the PMCA ATPase which are encoded by a multigene family.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATPase, Ca++ transporting, plasma membrane 1; ATPase, Ca++ transporting, plasma membrane 2; ATPase, Ca++ transporting, plasma membrane 3; ATPase, Ca++ transporting, plasma membrane 4; calcium pump; cilia and flagella associated protein 39; Matrix-remodeling-associated protein 1; plasma membrane Ca(2+)-ATPase; plasma membrane Ca2+ pump 2; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 1; Plasma membrane calcium ATPase isoform 2; Plasma membrane calcium ATPase isoform 3; Plasma membrane calcium ATPase isoform 4; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; Plasma membrane calcium pump isoform 2; Plasma membrane calcium pump isoform 3; Plasma membrane calcium pump isoform 4; plasma membrane calcium-ATPase isoform 1 alternative splice variant k at splice site B and splice variant b at splice site C; plasma membrane calcium-ATPase isoform 2 alternative splice variant a at splice site C; plasma membrane calcium-ATPase isoform 2, splice variant i; plasma membrane calcium-ATPase isoform 3, splice variant a at alternative splice site C; plasma membrane calcium-ATPase isoform 4 alternative splice variant x at splice site A; plasma membrane calcium-ATPase isoform 4, splice variant b at alternative splice site C; Plasma membrane calcium-transporting ATPase 1; Plasma membrane calcium-transporting ATPase 2; Plasma membrane calcium-transporting ATPase 3; Plasma membrane calcium-transporting ATPase 4; PMCA1; PMCA2; PMCA3; PMCA4; sarcolemmal calcium pump; unnamed protein product

          View more View less

          Gene Aliases: ATP2B1; ATP2B2; ATP2B3; ATP2B4; CFAP39; CLA2; DFNA82; MRD66; MXRA1; OPCA; PMCA1; PMCA1kb; PMCA2; PMCA2a; PMCA2i; PMCA3; PMCA3a; PMCA4; PMCA4b; PMCA4x; SCAX1

          View more View less

          UniProt ID: (Human) P20020, (Human) Q01814, (Human) Q16720, (Human) P23634

          View more View less

          Entrez Gene ID: (Human) 490, (Human) 491, (Human) 492, (Human) 493

          View more View less

          Function(s)
          nucleotide binding calcium-transporting ATPase activity protein binding calmodulin binding ATP binding calcium ion transmembrane transporter activity ATPase activity metal ion binding molecular function inhibitor activity calcium ion binding PDZ domain binding P-type calcium transporter activity involved in regulation of postsynaptic cytosolic calcium ion concentration P-type calcium transporter activity involved in regulation of presynaptic cytosolic calcium ion concentration sodium channel regulator activity protein kinase binding protein phosphatase 2B binding nitric-oxide synthase inhibitor activity nitric-oxide synthase binding scaffold protein binding
          Process(es)
          neural retina development regulation of transcription from RNA polymerase II promoter ion transport calcium ion transport cellular calcium ion homeostasis spermatogenesis negative regulation of gene expression urinary bladder smooth muscle contraction negative regulation of angiogenesis hippocampus development sperm motility ion transmembrane transport nitric oxide-cGMP-mediated signaling pathway negative regulation of blood vessel endothelial cell migration negative regulation of nitric oxide biosynthetic process regulation of cytosolic calcium ion concentration response to hydrostatic pressure calcium ion transmembrane transport negative regulation of calcineurin-NFAT signaling cascade cellular response to epinephrine stimulus negative regulation of adrenergic receptor signaling pathway calcium ion transmembrane import into cytosol calcium ion import across plasma membrane negative regulation of the force of heart contraction transport across blood-brain barrier negative regulation of arginine catabolic process calcium ion export regulation of sodium ion transmembrane transport negative regulation of cellular response to vascular endothelial growth factor stimulus regulation of cell cycle G1/S phase transition positive regulation of protein localization to plasma membrane negative regulation of cardiac muscle hypertrophy in response to stress negative regulation of citrulline biosynthetic process regulation of cardiac conduction cellular response to acetylcholine negative regulation of cytokine production regulation of vascular smooth muscle contraction regulation of blood pressure positive regulation of bone mineralization negative regulation of cytosolic calcium ion concentration positive regulation of calcium ion transport regulation of cellular response to insulin stimulus calcium ion export from cell sensory perception of sound neuron differentiation regulation of postsynaptic cytosolic calcium ion concentration regulation of presynaptic cytosolic calcium ion concentration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline