Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Nodal Antibodies

          Invitrogen

          Nodal Polyclonal Antibody

          View all (24) Nodal antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Nodal Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Nodal Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Nodal Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Nodal Antibody (PA5-118881) in WB

          Western blot analysis of Nodal in C6 cells using a Nodal polyclonal antibody (Product # PA5-118881). Observed band: 30 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Nodal Antibody in Western Blot (WB)
          Nodal Polyclonal Antibody

          Product Details

          PA5-118881

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human NODAL(Accession Q96S42), corresponding to amino acid residues L258-V308.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2903381

          Product Specific Information

          Antibody detects endogenous levels of total NODAL.

          Target Information

          NODAL is an important TGF-beta family protein in cellular differentiation. While constitutively expressed in many developing tissues, NODAL is problematically associated with the differentiation and development of many cancers. The expression of NODAL is regulated by a Notch signaling pathway, which when disrupted contributes to the aggressive behavior of metastatic cancer cells, particularly in melanoma. The abundance of NODAL expression in melanoma can help determine the stage of cancer and metastatic type/progression when tested in IHC. NODAL interactions with Cripto also predispose toward tumor progression. Overexpression of NODAL is also associated with preeclamptic placenta.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: early embryo mesoderm formation; MGC138230; Nodal; Nodal homolog; transgenic lethal 413.d

          View more View less

          Gene Aliases: HTX5; NODAL; Tg.413d

          View more View less

          UniProt ID: (Human) Q96S42, (Mouse) P43021

          View more View less

          Entrez Gene ID: (Human) 4838, (Rat) 294503, (Mouse) 18119

          View more View less

          Function(s)
          cytokine activity protein binding growth factor activity morphogen activity receptor agonist activity type I activin receptor binding transforming growth factor beta receptor binding growth factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development gastrulation with mouth forming second formation of primary germ layer mesoderm formation trophectodermal cell differentiation trophectodermal cellular morphogenesis neural fold formation liver development placenta development embryonic placenta development maternal placenta development vasculature development heart looping inhibition of neuroepithelial cell differentiation determination of left/right asymmetry in lateral mesoderm transmembrane receptor protein serine/threonine kinase signaling pathway transforming growth factor beta receptor signaling pathway germ cell development determination of left/right symmetry gastrulation nervous system development brain development endoderm development heart development female pregnancy cell proliferation positive regulation of cell proliferation embryonic pattern specification anterior/posterior axis specification anterior/posterior pattern specification regulation of signal transduction polarity specification of proximal/distal axis regulation of gastrulation positive regulation of vascular endothelial growth factor production negative regulation of cell development cell migration stem cell population maintenance positive regulation of cell-cell adhesion cell differentiation lung development floor plate morphogenesis somatic stem cell population maintenance embryonic heart tube development endodermal cell differentiation nodal signaling pathway cell migration involved in gastrulation cell fate commitment negative regulation of cell differentiation positive regulation of angiogenesis positive regulation of transcription from RNA polymerase II promoter axial mesodermal cell fate specification mesendoderm development digestive tract morphogenesis anatomical structure formation involved in morphogenesis embryonic cranial skeleton morphogenesis tissue morphogenesis formation of anatomical boundary positive regulation of epithelial cell proliferation digestive system development embryonic process involved in female pregnancy maternal process involved in parturition positive regulation of SMAD protein import into nucleus left lung morphogenesis negative regulation of androgen receptor signaling pathway epiblast cell-extraembryonic ectoderm cell signaling involved in anterior/posterior axis specification positive regulation of ERK1 and ERK2 cascade primitive streak formation apoptotic signaling pathway negative regulation of trophoblast cell migration negative regulation of chorionic trophoblast cell proliferation regulation of stem cell population maintenance negative regulation of reproductive process positive regulation of pathway-restricted SMAD protein phosphorylation positive regulation of activin receptor signaling pathway growth regulation of apoptotic process positive regulation of cysteine-type endopeptidase activity involved in apoptotic process regulation of MAPK cascade cell development positive regulation of sequence-specific DNA binding transcription factor activity SMAD protein signal transduction transforming growth factor beta receptor signaling pathway involved in primitive streak formation nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry positive regulation of receptor activity multicellular organism development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline