Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • OX40L (CD252) Antibodies

          Invitrogen

          OX40L Polyclonal Antibody

          View all (35) OX40L (CD252) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite OX40L Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          OX40L Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          OX40L Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OX40L Antibody (PA5-34516) in IHC

          Immunohistochemistry of TNFSF4 in human spleen tissue with OX40L Polyclonal Antibody (Product # PA5-34516) at 5 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OX40L Antibody in Immunohistochemistry (IHC)
          OX40L Antibody in Immunohistochemistry (IHC)
          OX40L Antibody in Western Blot (WB)
          OX40L Polyclonal Antibody

          Product Details

          PA5-34516

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-1 µg/mL
          -

          Immunohistochemistry (IHC)

          5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A 14 amino acid peptide near the carboxy terminus of human TNFSF4
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2551868

          Product Specific Information

          A suggested positive control is rat spleen tissue lysate.

          PA5-34516 can be used with blocking peptide PEP-1559.

          Target Information

          The tumor necrosis factor superfamily member TNFSF4 is a type II membrane bound, non-covalently linked homotrimeric protein. It is expressed on antigen presenting cells, such as dendritic cells and activated B-cells, and also on other cells such as vascular endothelial cells, mast cells, and natural killer cells. TNFSF4 signals specifically through the TNFRSF4 receptor, is expressed predominantly on CD4+T cells but also on certain activated CD8+T cells. TNFRSF4/TNFSF4 functions as a costimulatory signal, which is required for a productive interaction between antigen presenting cells and their target T-cells. It enhances cell proliferation and survival, and increases expression of RANTES, IL-2, IL-3, and IFNgamma. TNFRSF4/TNFSF4 signaling plays an important role in immunotolerance.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: atherosclerosis 1; CD134 ligand; CD252; Glycoprotein Gp34; OX 40 ligand/ gp 34; OX40 antigen ligand; OX40 ligand; OX40L; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; TNFSF4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor ligand 2B; Tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; unnamed protein product

          View more View less

          Gene Aliases: Ath-1; Ath1; CD134L; CD252; GP34; OX-40L; Ox40l; OX4OL; TNFSF4; TNLG2B; TXGP1; Txgp1l

          View more View less

          UniProt ID: (Human) P23510, (Rat) Q9Z2P3, (Mouse) P43488

          View more View less

          Entrez Gene ID: (Human) 7292, (Rat) 89814, (Mouse) 22164

          View more View less

          Function(s)
          receptor binding cytokine activity tumor necrosis factor receptor binding protein binding tumor necrosis factor receptor superfamily binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter positive regulation of cytokine production defense response to nematode acute inflammatory response positive regulation of immunoglobulin production positive regulation of T cell cytokine production regulation of adaptive immune response positive regulation of type 2 immune response positive regulation of immunoglobulin mediated immune response inflammatory response immune response cell communication signal transduction positive regulation of cell proliferation response to virus signaling negative regulation of interferon-gamma production negative regulation of interleukin-17 production positive regulation of chemokine production positive regulation of interferon-gamma production positive regulation of interleukin-10 production positive regulation of interleukin-12 production positive regulation of interleukin-13 production positive regulation of interleukin-4 production positive regulation of interleukin-6 production memory T cell activation T-helper 2 cell activation response to nitrogen dioxide positive regulation of T cell proliferation positive regulation of CD4-positive, alpha-beta T cell differentiation positive regulation of memory T cell differentiation negative regulation of regulatory T cell differentiation negative regulation of T-helper 1 cell differentiation positive regulation of T-helper 2 cell differentiation positive regulation of alpha-beta T cell proliferation regulation of inflammatory response positive regulation of inflammatory response positive regulation of B cell activation cellular response to lipopolysaccharide cellular response to prostaglandin E stimulus positive regulation of CD4-positive, alpha-beta T cell costimulation positive regulation of interleukin-4-dependent isotype switching to IgE isotypes cytokine production cholesterol metabolic process positive regulation of interleukin-2 production cellular response to nitrogen dioxide CD4-positive, alpha-beta T cell costimulation T cell proliferation positive regulation of activated T cell proliferation negative regulation of sequence-specific DNA binding transcription factor activity negative regulation of transcription, DNA-templated negative regulation of cytokine secretion positive regulation of immunoglobulin secretion chemokine (C-C motif) ligand 11 production positive regulation of T cell costimulation positive regulation of memory T cell activation positive regulation of T-helper 2 cell activation blood vessel development positive regulation of immune effector process innate immune response negative regulation of T cell apoptotic process negative regulation of activation-induced cell death of T cells positive regulation of T cell migration negative regulation of extrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline