Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • DST Antibodies

          Invitrogen

          DST Polyclonal Antibody

          View all (5) DST antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DST Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          DST Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DST Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DST Antibody (PA5-96034) in WB

          Western blot analysis of DST in extracts of various cell lines. Samples were incubated with DST Polyclonal antibody (Product # PA5-96034) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 90s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DST Antibody in Western Blot (WB)
          DST Polyclonal Antibody

          Product Details

          PA5-96034

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.97 mg/mL

          Amount

          97 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2807836

          Product Specific Information

          Immunogen sequence: MHSSSYSYRS SDSVFSNTTS TRTSLDSNEN LLLVHCGPTL INSCISFGSE SFDGHRLEML QQIANRVQRD SVICEDKLIL AGNALQSDSK RLESGVQFQN EAEIAGYILE CENLLRQHVI DVQILIDGKY YQADQLVQRV AKLRDEIMAL RNECSSVYSK GRILTTEQTK LMISGITQSL NSGFAQTLHP SLTSGLTQSL TPSLTSSSMT SGLSSGMTSR LTPSVTPAYT PGFPSGLVPN FSSGVEPNSL QTLKLMQIRK PLLKSSLLDQ; Positive Samples: THP-1, Rat kidney; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasm, Endoplasmic reticulum membrane, H zone, Lipid-anchor, Membrane, Nucleus, Nucleus envelope, Single-pass membrane protein, Z line, axon, cell cortex, cytoskeleton, cytoskeleton, hemidesmosome, myofibril, sarcomere

          Target Information

          As basal cells of stratified squamous epithelia begin to migrate in response to wound healing, they lose their cell-substrate adhesion junctions, the hemidesmosomes. The hemidesmosome is an adhesion structure of the epidermal-dermal junction in keratinocytes. When keratinocytes migrate laterally or upward to differentiate they must control the formation and disintegration of the hemidesmosomes. The bullous pemphigoid antigen BPAG1 is a hemidesmosomal protein of the cutaneous basement membrane zone. The primary sequence deduced from full-length human cDNAs predicts that this molecule consists of a central rod region and flanking globular domains. A neuronal isoform, BPAG1n3 is the result of differential splicing of BPAG1. BPAG1n3 is distinguished by its initial 32 amino acid residues and by the absence of the amino-terminal half of the actin-binding domain.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 230 kDa bullous pemphigoid antigen; 230/240 kDa bullous pemphigoid antigen; BPA; BPAG1eA; MACF-like product; BPAG1eB; second IF-binding domain product; bullous pemphigoid antigen; Bullous pemphigoid antigen 1; bullous pemphigoid antigen 1, isoform 5; bullous pemphigoid antigen 1, isoforms 1/2/3/4; bullous pemphigoid antigen 1, isoforms 6/7; Dystonia musculorum protein; Dystonin; Hemidesmosomal plaque protein; Microtubule actin cross-linking factor 2; trabeculin-beta; unnamed protein product

          View more View less

          Gene Aliases: 2310001O04Rik; A830042E19Rik; ah; athetoid; AW554249; BP230; BP240; BPA; Bpag; BPAG1; BPAG1-n; CATX-15; CATX15; D6S1101; DMH; DST; DT; EBS3; EBSB2; HSAN6; KIAA0728; MACF2; mKIAA0728; nmf203; nmf339

          View more View less

          UniProt ID: (Human) Q03001, (Mouse) Q91ZU6

          View more View less

          Entrez Gene ID: (Human) 667, (Mouse) 13518, (Rat) 316313

          View more View less

          Function(s)
          actin binding integrin binding structural molecule activity calcium ion binding protein binding microtubule binding protein homodimerization activity metal ion binding microtubule plus-end binding cytoskeletal protein binding protein C-terminus binding intermediate filament binding protein
          Process(es)
          microtubule cytoskeleton organization cytoskeleton organization cell adhesion integrin-mediated signaling pathway retrograde axonal transport response to wounding maintenance of cell polarity hemidesmosome assembly wound healing intermediate filament cytoskeleton organization cell motility cell cycle arrest axonogenesis regulation of microtubule polymerization or depolymerization cytoplasmic microtubule organization intracellular transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline