Search
Search

Entrez Gene ID: | 3420 |
Gene Name: | isocitrate dehydrogenase 3 (NAD(+)) beta |
Gene Aliases: |
RP46 |
Location: |
Chr.20:2658390-2664223 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDH3B | NM_006899.4 | NP_008830.2 | ||
| NM_174855.3 | NP_777280.1 | |||
| NR_136344.1 | ||||
| XM_005260716.1 | XP_005260773.1 | |||
| AK001905.1 | BAA91971.1 | |||
| AL050094.1 | CAB43266.1 | |||
| BC001960.1 | AAH01960.1 | |||
| BX119223.1 | ||||
| BX647393.1 |
Entrez Gene ID: | 10528 |
Gene Name: | NOP56 ribonucleoprotein |
Gene Aliases: |
NOL5A, SCA36 |
Location: |
Chr.20:2652532-2658393 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOP56 | NM_006392.3 | NP_006383.2 | ||
| NR_027700.2 | ||||
| AK097186.1 | ||||
| AK292721.1 | ||||
| BC004937.1 | AAH04937.1 | |||
| BX641071.1 | ||||
| BX647439.1 | ||||
| BX647513.1 | ||||
| Y12065.1 | CAA72789.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057464 | Chr.20:2650623 - 2989686 on Build GRCh38 | Gain |
|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
| esv3644998 | Chr.20:2148722 - 2690694 on Build GRCh38 | Gain |
|
| nsv585266 | Chr.20:2657933 - 2704278 on Build GRCh38 | Loss |
|
Set Membership: |
|