Search
Search

Entrez Gene ID: | 8676 |
Gene Name: | syntaxin 11 |
Gene Aliases: |
FHL4, HLH4, HPLH4 |
Location: |
Chr.6:144140044-144191939 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX11 | NM_003764.3 | 2 | 405 | NP_003755.2 |
| XM_011536213.2 | 3 | 517 | XP_011534515.1 | |
| XM_011536214.2 | 3 | 721 | XP_011534516.1 | |
| XM_011536217.2 | 2 | 242 | XP_011534519.1 | |
| XM_011536218.2 | 2 | 328 | XP_011534520.1 | |
| XM_017011400.1 | 2 | 384 | XP_016866889.1 | |
| AF038898.1 | 1 | 213 | AAD02107.1 | |
| AF044309.1 | 2 | 396 | AAC24031.1 | |
| AF071504.1 | 2 | 267 | AAC24004.1 | |
| BC033519.1 | 2 | 390 | AAH33519.1 | |
| BM150302.1 | 2 | 406 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830831 | Chr.6:144180234 - 144328373 on Build GRCh38 | Gain |
|
Set Membership: |
|