Search
Search

Entrez Gene ID: | 23759 |
Gene Name: | peptidylprolyl isomerase like 2 |
Gene Aliases: |
CYC4, CYP60, Cyp-60, UBOX7, hCyP-60 |
Location: |
Chr.22:21665984-21698276 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIL2 | NM_001317996.1 | 21 | 2084 | NP_001304925.1 |
| NM_014337.3 | 20 | 2454 | NP_055152.1 | |
| NM_148175.2 | NP_680480.1 | |||
| NM_148176.2 | NP_680481.1 | |||
| XM_005261448.3 | XP_005261505.1 | |||
| XM_011530041.2 | 21 | 2080 | XP_011528343.1 | |
| XM_011530042.2 | XP_011528344.1 | |||
| XM_011530043.2 | XP_011528345.1 | |||
| XM_011530044.2 | XP_011528346.1 | |||
| XM_011530046.2 | XP_011528348.1 | |||
| XM_011530047.2 | XP_011528349.1 | |||
| XM_011530049.2 | 22 | 2130 | XP_011528351.1 | |
| XM_011530050.2 | 20 | 1993 | XP_011528352.1 | |
| XM_011530051.2 | 21 | 2020 | XP_011528353.1 | |
| XM_017028705.1 | 12 | 1326 | XP_016884194.1 | |
| XM_017028706.1 | XP_016884195.1 | |||
| AK097249.1 | ||||
| AK127259.1 | 22 | 2566 | ||
| AK299206.1 | ||||
| BC000022.2 | AAH00022.1 | |||
| BC028385.2 | 21 | 1864 | AAH28385.1 | |
| CR456548.1 | CAG30434.1 | |||
| U37219.1 | 20 | 2430 | AAC50376.1 | |
| U37220.1 | AAC50377.1 | |||
| U37221.1 | AAC50378.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1293e212 | Chr.22:21685588 - 21699487 on Build GRCh38 | Loss |
|
| nsv510804 | Chr.22:21670242 - 21778475 on Build GRCh38 | Deletion |
|
Set Membership: |
|