Search
Search

Entrez Gene ID: | 5071 |
Gene Name: | parkin RBR E3 ubiquitin protein ligase |
Gene Aliases: |
AR-JP, LPRS2, PDJ, PRKN |
Location: |
Chr.6:161347417-162727802 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARK2 | NM_004562.2 | NP_004553.2 | ||
| NM_013987.2 | NP_054642.2 | |||
| NM_013988.2 | NP_054643.2 | |||
| XM_011535863.1 | XP_011534165.1 | |||
| XM_017010908.1 | XP_016866397.1 | |||
| XM_017010909.1 | XP_016866398.1 | |||
| AB009973.1 | BAA25751.1 | |||
| AB245403.1 | ||||
| AF381282.1 | AAM21457.1 | |||
| AF381283.1 | AAM21458.1 | |||
| AF381284.1 | AAM21459.1 | |||
| AK292590.1 | ||||
| BC022014.2 | AAH22014.1 | |||
| EF375726.1 | ||||
| GU345837.1 | ||||
| GU345838.1 | ||||
| GU345839.1 | ||||
| GU345840.1 | ||||
| GU357501.1 | ||||
| GU357502.1 | ||||
| GU361466.1 | ||||
| GU361467.1 | ||||
| GU361468.1 | ||||
| GU361469.1 | ||||
| GU361470.1 | ||||
| GU361471.1 | ||||
| KC357595.1 | ||||
| KC774171.1 | ||||
| KJ534911.1 | ||||
| KJ534912.1 | ||||
| KU178240.1 | ||||
| KU178241.1 | ||||
| KU178242.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018409 | Chr.6:161809542 - 162125672 on Build GRCh38 | Loss |
|
| nsv1026147 | Chr.6:161704721 - 162260681 on Build GRCh38 | Loss |
|
| esv3611465 | Chr.6:161567774 - 162085535 on Build GRCh38 | Gain |
|
| nsv527260 | Chr.6:161950673 - 162054402 on Build GRCh38 | Loss |
|
| nsv523485 | Chr.6:161788587 - 162333574 on Build GRCh38 | Loss |
|
| dgv6173n100 | Chr.6:161948825 - 162054024 on Build GRCh38 | Loss |
|
| nsv5580 | Chr.6:161952895 - 161982113 on Build GRCh38 | Deletion |
|
| esv3571506 | Chr.6:161948824 - 162058246 on Build GRCh38 | Loss |
|
| nsv1029749 | Chr.6:161543443 - 162045027 on Build GRCh38 | Loss |
|
| nsv518931 | Chr.6:161372287 - 161985303 on Build GRCh38 | Gain |
|
| esv2752093 | Chr.6:161900345 - 162288916 on Build GRCh38 | Gain |
|
| dgv6169n100 | Chr.6:161644335 - 161956796 on Build GRCh38 | Loss |
|
| nsv605074 | Chr.6:161934723 - 161995173 on Build GRCh38 | Gain |
|
| dgv6172n100 | Chr.6:161925250 - 162180183 on Build GRCh38 | Loss |
|
| esv3571505 | Chr.6:161937230 - 162847872 on Build GRCh38 | Loss |
|
| nsv526031 | Chr.6:161907241 - 162333574 on Build GRCh38 | Gain |
|
| esv2759486 | Chr.6:161769724 - 161990218 on Build GRCh38 | Gain |
|
| nsv5579 | Chr.6:161940654 - 161975142 on Build GRCh38 | Insertion |
|
| esv3611478 | Chr.6:161853183 - 162068074 on Build GRCh38 | Gain |
|
Set Membership: |
|