Search
Search

Entrez Gene ID: | 8559 |
Gene Name: | pre-mRNA processing factor 18 |
Gene Aliases: |
PRP18, hPrp18 |
Location: |
Chr.10:13586811-13645279 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF18 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759734 | Chr.10:13636670 - 13644642 on Build GRCh38 | Loss |
|
| nsv831790 | Chr.10:13536081 - 13716895 on Build GRCh38 | Loss |
|
| esv3891761 | Chr.10:13544627 - 13695972 on Build GRCh38 | Gain |
|
| esv2761575 | Chr.10:13581531 - 13659502 on Build GRCh38 | Gain |
|