Search
Search

Entrez Gene ID: | 10267 |
Gene Name: | receptor activity modifying protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:237858880-237912106 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAMP1 | NM_001308353.2 | NP_001295282.1 | ||
| NM_005855.4 | NP_005846.1 | |||
| XM_017003152.3 | XP_016858641.1 | |||
| XM_017003153.3 | XP_016858642.1 | |||
| XM_017003156.3 | XP_016858645.1 | |||
| AI017108.1 | ||||
| AJ001014.1 | CAA04472.1 | |||
| AY265457.1 | AAP23298.1 | |||
| BC000548.2 | AAH00548.1 | |||
| BI548986.1 | ||||
| BM978161.1 | ||||
| CR542032.1 | CAG46829.1 | |||
| CR542044.1 | CAG46841.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4075099 | Chr.2:237888275 - 237918497 on Build GRCh38 | Duplication |
|
| nsv508205 | Chr.2:237889539 - 237970025 on Build GRCh38 | Deletion |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
| nsv6337133 | Chr.2:237890456 - 237925443 on Build GRCh38 | Duplication |
|
Set Membership: |
|