Search
Search

Entrez Gene ID: | 4288 |
Gene Name: | marker of proliferation Ki-67 |
Gene Aliases: |
KIA, MIB-, MIB-1, PPP1R105 |
Location: |
Chr.10:128096659-128126405 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MKI67 | NM_001145966.1 | NP_001139438.1 | ||
| NM_002417.4 | NP_002408.3 | |||
| XM_006717864.3 | XP_006717927.2 | |||
| XM_011539818.2 | XP_011538120.1 | |||
| AK299254.1 | ||||
| X65550.1 | CAA46519.1 | |||
| X65551.1 | CAA46520.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv52n21 | Chr.10:128091905 - 128109225 on Build GRCh38 | Loss |
|
| esv34105 | Chr.10:128072029 - 128563717 on Build GRCh38 | Loss |
|
| dgv989n100 | Chr.10:128040003 - 128445808 on Build GRCh38 | Gain |
|
Set Membership: |
|