Search
Search

Entrez Gene ID: | 1288 |
Gene Name: | collagen type IV alpha 6 chain |
Gene Aliases: |
CXDELq22.3, DELXq22.3, DFNX6 |
Location: |
Chr.X:108155607-108439497 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL4A6 | NM_001287758.1 | NP_001274687.1 | ||
| NM_001287759.1 | NP_001274688.1 | |||
| NM_001287760.1 | NP_001274689.1 | |||
| NM_001847.3 | NP_001838.2 | |||
| NM_033641.3 | NP_378667.1 | |||
| XM_006724617.3 | XP_006724680.1 | |||
| XM_011530852.2 | XP_011529154.1 | |||
| XM_011530854.2 | XP_011529156.1 | |||
| BC005305.1 | AAH05305.1 | |||
| BC070203.1 | AAH70203.1 | |||
| BC140855.1 | ||||
| BC144669.1 | ||||
| BT007228.1 | AAP35892.1 | |||
| CN313347.1 | ||||
| D21337.1 | BAA04809.1 | |||
| L22763.1 | AAA16338.1 | |||
| U04845.1 | AAA19569.2 |
Set Membership: |
|