Search
Search

Entrez Gene ID: | 101927891 |
Gene Name: | PKN2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:88537513-88685204 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKN2-AS1 | NR_110682.1 | |||
| AK123834.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546772 | Chr.1:88017880 - 88579655 on Build GRCh38 | Loss |
|
| nsv1002331 | Chr.1:88493843 - 88584211 on Build GRCh38 | Loss |
|
| nsv546773 | Chr.1:88320368 - 88696449 on Build GRCh38 | Loss |
|
| nsv470725 | Chr.1:87987620 - 88579655 on Build GRCh38 | Loss |
|