Search
Search

Entrez Gene ID: | 5290 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha |
Gene Aliases: |
CLOVE, CWS5, MCAP, MCM, MCMTC, PI3K, PI3K-alpha, p110-alpha |
Location: |
Chr.3:179148114-179240093 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CA | NM_006218.3 | NP_006209.2 | ||
| XM_006713658.3 | XP_006713721.1 | |||
| XM_011512894.2 | XP_011511196.1 | |||
| AB210020.1 | ||||
| AK292940.1 | ||||
| AK297203.1 | ||||
| AK302608.1 | ||||
| BC113601.1 | ||||
| BC113603.1 | ||||
| U79143.1 | AAB39753.1 | |||
| Z29090.1 | CAA82333.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893827 | Chr.3:179207079 - 179304740 on Build GRCh38 | Gain |
|
| dgv4999n100 | Chr.3:179227018 - 179528391 on Build GRCh38 | Gain |
|
Set Membership: |
|