Search
Search

Entrez Gene ID: | 13599 |
Gene Name: | endothelin converting enzyme-like 1 |
Gene Aliases: |
DINE, xce |
Location: |
Chr.1:87142874-87157085 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ecel1 | NM_001277925.1 | NP_001264854.1 | ||
| NM_021306.3 | NP_067281.2 | |||
| AB026294.1 | BAA95005.1 | |||
| AK039467.1 | ||||
| AK159365.1 | BAE35023.1 | |||
| BC057569.1 | AAH57569.1 |
Set Membership: |
|