Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: tre-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306844.3 | NP_001293773.1 | 7 - 8 | 867 | 99 | |
NM_001380293.1 | NP_001368400.1 | 6 - 7 | 726 | 99 | ||
NM_068657.7 | NP_501058.2 | 9 - 10 | 1125 | 99 | ||
GenBank mRNA | AJ512334.1 | - | 8 - 9 | 1102 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |