Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Actl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500750.2 | XP_003500798.1 | 1 | 1170 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Both primers and probe map within a single exon |