Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GlcAT-P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259784.2 | NP_001246713.1 | 5 - 6 | 1442 | 72 | |
NM_140191.3 | NP_648448.1 | 5 - 6 | 1543 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |