Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: pvalb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205573.1 | NP_991136.1 | 4 - 5 | 516 | 85 | |
XM_021469917.1 | XP_021325592.1 | 4 - 5 | 348 | 85 | ||
GenBank mRNA | AF467913.1 | - | 4 - 5 | 516 | 85 | |
BC081523.1 | - | 4 - 5 | 534 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |