Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: shox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126411.1 | NP_001119883.1 | 2 - 3 | 578 | 79 | |
XM_005167854.4 | XP_005167911.1 | 2 - 3 | 604 | 79 | ||
GenBank mRNA | BC107618.1 | - | 2 - 3 | 589 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |