Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03462137_g1
          See other LOC100128076 GE Assays ›
          Gene Symbol:
          LOC100128076
          Gene Name:
          protein tyrosine phosphatase pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 92132834 - 92138629 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03462137_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100128076
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015444.1 11 - 12 1148 75
          GenBank mRNA AY786555.1 - 11 - 12 1148 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream serine palmitoyltransferase long chain base subunit 1 SPTLC1 HSAN1,
          HSN1,
          LBC1,
          LCB1,
          SPT1,
          SPTI
          -21.8kb Anti-sense Chr. 9 - 92031134 - 92115474 on NCBI Build GRCh38
          Downstream long intergenic non-protein coding RNA 475 LINC00475 C9orf44 3.3kb Sense Chr. 9 - 92141467 - 92159608 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900007945.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000433.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16239868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06818026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27158529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27158518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06818017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16239865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16239907 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27158535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06818023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06818021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27158520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16239873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27158517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06818018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16239882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27158521 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06818022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16239860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06818020 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 921051 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 202546 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 230991 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 143968 BC117698.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 143968 BX648940.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 143968 CR749794.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 143968 NM_001007525.1 refseq_mrna_human_NM
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4398 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4397 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P16919090 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3309481 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239907 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158521 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818023 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239865 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158529 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158503 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818021 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239858 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239907 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158503 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158529 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818018 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818017 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818026 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239865 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158517 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27158521 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16239882 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06818023 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P02132424 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818026 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818031 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818030 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16239910 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16239959 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818023 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818019 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818012 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16239987 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16239943 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818015 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818035 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818043 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818040 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818017 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16239947 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818020 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818016 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818033 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818044 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818022 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818029 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06818014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09004572.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09004572.hg.1 TC09000433.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09004573.hg.1 NR_015444.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09004573.hg.1 TC09000433.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900153462.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900153462.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900153462.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900153462.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900153462.hg.1 TC0900007945.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007945.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900153464.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900153464.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900153464.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900153464.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900153464.hg.1 TC0900007945.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900153463.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900153463.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900153463.hg.1 TC0900007945.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900153466.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900153466.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900153466.hg.1 ENST00000412768.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900153466.hg.1 NR_015444.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900153466.hg.1 TC0900007945.hg.1 Clariom D TC
          Human Genome U95 Set 81691_at - -
          Human Genome U95 Set 81691_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038934 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A038934 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 137733 - -
          ABI Human Genome Survey Microarray Version 2.0 192445 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline