Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RPS10P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026667.1 | 1 | 61 | 151 | ||
GenBank mRNA | AK055590.1 | - | 1 | 1259 | 151 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cysteine and glycine rich protein 1 | CSRP1 |
CRP, CRP1, CSRP, CYRP, D1S181E, HEL-141, HEL-S-286 |
-12.6kb | Anti-sense | Chr. 1 - 201483530 - 201507259 on NCBI Build GRCh38 |
Downstream | neuron navigator 1 | NAV1 |
POMFIL3, STEERIN1, UNC53H1 |
18.5kb | Sense | Chr. 1 - 201648322 - 201826974 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728839 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789012 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728829 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728834 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789068 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789043 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728848 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728860 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728836 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728821 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728817 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728837 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728842 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728843 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728818 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728832 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728830 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728800 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728819 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728867 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728849 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789036 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789050 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728811 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789085 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728810 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728859 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728841 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728850 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728815 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728861 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789057 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728868 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789053 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728804 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789041 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15161844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728864 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15161845 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15161831 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728812 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20789023 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728822 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728809 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728845 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728862 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728851 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00728857 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 52318 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 808168 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 921733 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 835819 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 747637 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P203013 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P203013 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 42299 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 45036 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P203013 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P203013 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20789011 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20789006 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20788999 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728781 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20789005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728785 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15161824 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20789005 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728781 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728785 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20789006 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15161824 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20788999 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00728783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20789011 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P203013 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15411012 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 172180 | BC003649.2 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 172180 | BC012297.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 172180 | BC021577.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 172180 | BC033658.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 172180 | BC040446.2 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 172180 | BC053847.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 172180 | CR591718.1 | genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 172180 | NM_001810.5 | refseq_mrna_human_NM |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15411011 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00232833 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P18017954 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15411012 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35428282 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P203017 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728767 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728769 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728791 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728794 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728784 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728775 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728766 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728788 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728783 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728777 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728800 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728772 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728798 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728780 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00728796 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15411012 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35428282 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7908693 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7908692 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1566720_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01025673.hg.1 | TC01001660.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01025673.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01025673.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | ENST00000482594.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018336.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016061.hg.1 | ENST00000415019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016061.hg.1 | ENST00000415019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016061.hg.1 | ENST00000439139.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007347.hg.1 | ENST00000468490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300009222.hg.1 | ENST00000430654.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007347.hg.1 | ENST00000468490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300009222.hg.1 | ENST00000430654.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240196.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240196.hg.1 | ENST00000482594.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240196.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240196.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240196.hg.1 | ENST00000482594.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240196.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240196.hg.1 | TC0100018336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | ENST00000482594.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018336.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240195.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240195.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240195.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240195.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240195.hg.1 | TC0100018336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100240197.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240197.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240197.hg.1 | ENST00000441932.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100240197.hg.1 | NR_026667.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100240197.hg.1 | TC0100018336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100016061.hg.1 | ENST00000415019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016061.hg.1 | ENST00000415019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016061.hg.1 | ENST00000439139.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 217336_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 217336_at | - | - |
Human Genome U133 Plus 2.0 Array | 1566720_at | - | - |
Human Genome U133 Plus 2.0 Array | 1566720_PM_at | - | - |
Human Genome U133A 2.0 Array | 217336_at | - | - |
Human Genome U133A 2.0 Array | 217336_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566720_at10 | - | - |
Human Genome U133 Set | 217336_at | - | - |
Human Genome U133 Set | 217336_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036072 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037741 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037741 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036072 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1805348 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1805348 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1805348 | - | - |