Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vti1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016800.3 | NP_058080.2 | 3 - 4 | 491 | 84 | |
GenBank mRNA | AF035208.1 | - | 3 - 4 | 378 | 84 | |
AF035821.1 | - | 3 - 4 | 364 | 84 | ||
AK160624.1 | - | 2 - 3 | 425 | 84 | ||
BC009810.1 | - | 3 - 4 | 421 | 4220215 | 84 | |
BC080204.1 | - | 3 - 4 | 467 | 3672445 | 84 | |
CX239415.1 | - | 3 - 4 | 468 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |