Search
Search

Gata1| Gene Symbol: Gata1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008089.2 | NP_032115.1 | 2 - 3 | 380 | 76 | |
| GenBank mRNA | AK146915.1 | - | 2 - 3 | 365 | 76 | |
| BC052653.1 | - | 2 - 3 | 347 | 30039400 | 76 | |
| X15763.1 | - | 2 - 3 | 285 | 76 | ||
Set Membership: |
|