Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Grid2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159321.1 | NP_001152793.1 | 15 - 16 | 2896 | 149 | |
NM_133355.1 | NP_579933.1 | 14 - 15 | 2200 | 149 | ||
GenBank mRNA | AF099933.2 | - | 14 - 15 | 2200 | 149 | |
BC075624.1 | - | 2252 | 6841572 | 149 | ||
DQ193535.1 | - | 15 - 16 | 2896 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |