Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prame | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029459.2 | NP_083735.2 | 6 - 7 | 603 | 109 | |
GenBank mRNA | AK015971.1 | - | 6 - 7 | 604 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |