Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024762.1 | NP_001019933.1 | 6 - 7 | 663 | 71 | |
XM_006244686.2 | XP_006244748.1 | 4 - 5 | 492 | 71 | ||
XM_006244689.2 | XP_006244751.1 | 4 - 5 | 443 | 71 | ||
XM_006244690.2 | XP_006244752.1 | 4 - 5 | 509 | 71 | ||
XM_006244691.1 | XP_006244753.1 | 4 - 5 | 510 | 71 | ||
GenBank mRNA | AJ011607.1 | - | 3 - 4 | 347 | 71 | |
BC097454.1 | - | 6 - 7 | 663 | 7456950 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |