Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tulp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763338.1 | XP_008761560.1 | 1 - 2 | 108 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |