Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sarm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105817.1 | NP_001099287.1 | 5 - 6 | 1759 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |