Search
Search

betaTub97EF| Gene Symbol: betaTub97EF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170282.2 | NP_001163753.1 | 1 - 2 | 286 | 55 | |
| NM_143349.2 | NP_651606.2 | 1 - 2 | 286 | 55 | ||
| GenBank mRNA | AY118611.1 | - | 1 - 2 | 269 | 55 | |
Set Membership: |
|