Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm03419415_m1
          See other RL GE Assays ›
          Gene Symbol:
          rl
          Gene Name:
          rolled
          Gene Aliases:
          12559, BcDNA:RE08694, CG12559, CG18732, CT34260, CT39192, D-ERK, DERK, DERK-A, DmERK, DmERK-A, DmERKA, DmErk, DmMAPK, Dmel\CG12559, Dmel_CG12559, DpERK, DpErk, Dsor2, E(sina)7, EC2-1, EK2-1, ERK, ERK-A, ERK1/2, ERKA, ERKa, EY2-2, Erk, Erk/Map kinase, Erk1, Erk1/2, ErkA, GroupII, MAP-k, MAPK, MapK, Mapk, RL, Rl, SEM, SR2-1, Sem, Su(Raf)2B, dERK, dErk, dm-dpERK, dp-ERK, dpERK, dpERK1, dpERk, dpErk, dpMAPK, erk, l(2)41Ac, l(2R)EMS45-39, mapk, p-ERK, pERK, pMAPK, pMapK, rl/ERK, rl/MAPK, rll, sem
          Chromosome Location:
          Chr.2R: 1071462 - 1125927 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0003256
          Assay ID Dm03419415_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: rl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001015121.3 NP_001015121.3 6 - 7 731 88
          NM_001015122.3 NP_001015122.1 5 - 6 662 88
          NM_001015123.3 NP_001015123.1 4 - 5 843 88
          NM_001110878.4 NP_001104348.2 6 - 7 700 88
          NM_001110879.2 NP_001104349.1 5 - 6 621 88
          NM_001300706.2 NP_001287635.1 5 - 6 638 88
          GenBank mRNA AY070996.1 - 5 - 6 662 88
          M95124.1 - 4 - 5 788 88

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          mitotic cell cycle
          cell surface receptor signaling pathway
          epidermal growth factor receptor signaling pathway
          terminal region determination
          terminal branching, open tracheal system
          R7 cell fate commitment
          imaginal disc-derived wing vein specification
          imaginal disc-derived wing morphogenesis
          leg disc proximal/distal pattern formation
          metamorphosis
          positive regulation of cell proliferation
          insulin receptor signaling pathway
          torso signaling pathway
          determination of adult lifespan
          fibroblast growth factor receptor signaling pathway
          cellular response to starvation
          regulation of gene expression
          negative regulation of macroautophagy
          cellular response to reactive oxygen species
          lymph gland plasmatocyte differentiation
          lymph gland crystal cell differentiation
          tracheal pit formation in open tracheal system
          intracellular signal transduction
          sevenless signaling pathway
          positive regulation of cell size
          positive regulation of photoreceptor cell differentiation
          dorsal appendage formation
          vascular endothelial growth factor receptor signaling pathway
          behavioral response to ethanol
          modulation of synaptic transmission
          positive regulation of sequence-specific DNA binding transcription factor activity
          defense response to virus
          branching involved in open tracheal system development
          ERK1 and ERK2 cascade
          cellular response to arsenic-containing substance
          cellular response to cadmium ion
          positive regulation of wound healing
          regulation of heart morphogenesis
          regulation of response to drug
          protein serine/threonine kinase activity
          MAP kinase activity
          ATP binding
          transcription factor binding
          protein kinase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0