Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: sh3gl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105606.1 | NP_001099076.1 | 5 - 6 | 67 | ||
NM_200301.1 | NP_956595.1 | 5 - 6 | 601 | 67 | ||
XM_005168418.1 | XP_005168475.1 | 5 - 6 | 67 | |||
GenBank mRNA | BC049513.1 | - | 5 - 6 | 601 | 67 | |
BC153638.1 | - | 5 - 6 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |