Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Gg03315081_m1
          See other ZNF644 GE Assays ›
          Gene Symbol:
          ZNF644
          Gene Name:
          zinc finger protein 644
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 14981283 - 15059203 on Build GRCg6a
          Species:
          Chicken
          Species Specific ID (Flybase ID):
          -
          Assay ID Gg03315081_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZNF644
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_001232675.4 XP_001232676.1 6 - 7 2936 83
          XM_015290666.2 XP_015146152.1 9 - 10 3967 83
          XM_015290667.2 XP_015146153.1 11 - 12 3953 83
          XM_015290668.2 XP_015146154.1 9 - 10 3788 83
          XM_015290669.2 XP_015146155.1 10 - 11 4063 83
          XM_015290670.2 XP_015146156.1 10 - 11 3883 83
          XM_015290671.2 XP_015146157.1 9 - 10 5042 83
          XM_015290672.2 XP_015146158.1 10 - 11 4015 83
          XM_015290673.2 XP_015146159.1 9 - 10 3729 83
          XM_015290674.2 XP_015146160.1 9 - 10 3659 83
          XM_015290677.2 XP_015146163.1 8 - 9 3910 83
          XM_015290678.2 XP_015146164.1 8 - 9 3730 83
          XM_015290680.2 XP_015146166.1 8 - 9 3828 83
          XM_015290681.2 XP_015146167.1 7 - 8 3264 83
          XM_015290683.2 XP_015146169.1 6 - 7 3170 83
          XM_015290684.2 XP_015146170.1 5 - 6 3113 83
          XM_025153002.1 XP_025008770.1 11 - 12 4133 83
          XM_025153003.1 XP_025008771.1 10 - 11 3835 83
          XM_025153004.1 XP_025008772.1 8 - 9 3665 83
          XM_025153005.1 XP_025008773.1 9 - 10 3771 83
          XM_025153006.1 XP_025008774.1 8 - 9 3713 83
          XM_025153007.1 XP_025008775.1 8 - 9 3693 83
          XM_025153008.1 XP_025008776.1 7 - 8 3604 83
          XM_025153009.1 XP_025008777.1 7 - 8 3694 83
          XM_025153010.1 XP_025008778.1 7 - 8 3542 83
          XM_025153011.1 XP_025008779.1 5 - 6 2932 83
          GenBank mRNA AJ720546.1 - 6 - 7 2901 83

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline