Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025155349.1 | XP_025011117.1 | 3 - 4 | 273 | 79 | |
XM_025155350.1 | XP_025011118.1 | 3 - 4 | 306 | 79 | ||
GenBank mRNA | BU237198.1 | - | 3 - 4 | 79 | ||
CN218549.1 | - | 3 - 4 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |