Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: P3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001530.1 | NP_001001530.1 | 10 - 11 | 1501 | 98 | |
XM_015291384.2 | XP_015146870.1 | 10 - 11 | 1585 | 98 | ||
XM_015291385.2 | XP_015146871.1 | 10 - 11 | 1192 | 98 | ||
GenBank mRNA | AY463529.1 | - | 10 - 11 | 1501 | 98 | |
CR733175.1 | - | 8 - 9 | 747 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |