Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00169233_m1
          See other AHR GE Assays ›
          Gene Symbol:
          AHR
          Gene Name:
          aryl hydrocarbon receptor
          Gene Aliases:
          FVH3, RP85, bHLHe76
          Chromosome Location:
          Chr.7: 17298652 - 17346147 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00169233_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AHR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001621.5 NP_001612.1 6 - 7 1318 105
          GenBank mRNA AK307495.1 - 6 - 7 1318 105
          BC069390.1 - 6 - 7 775 105
          BC070080.1 - 6 - 7 1236 105
          D16354.1 - 6 - 7 1287 105
          L19872.1 - 6 - 7 1080 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700006795.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000112.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Human Nuclear Receptors HUMAN AHR PATHWAY Inventoried Human_Nuclear_Receptors Human_AHR_pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AHR

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          regulation of adaptive immune response
          negative regulation of T cell mediated immune response to tumor cell
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          xenobiotic metabolic process
          apoptotic process
          response to xenobiotic stimulus
          response to toxic substance
          regulation of gene expression
          intracellular receptor signaling pathway
          regulation of B cell proliferation
          circadian regulation of gene expression
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          negative regulation of inflammatory response
          intestinal epithelial structure maintenance
          cellular response to molecule of bacterial origin
          cellular response to cAMP
          cellular response to forskolin
          cellular response to 2,3,7,8-tetrachlorodibenzodioxine
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter proximal region sequence-specific DNA binding
          TFIID-class transcription factor binding
          transcription coactivator binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          TBP-class protein binding
          protein homodimerization activity
          protein heterodimerization activity
          protein dimerization activity
          Hsp90 protein binding
          RNA polymerase II sequence-specific DNA binding transcription factor binding
          E-box binding
          sequence-specific double-stranded DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000242057.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000463496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000475440.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000481944.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000481944.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000481944.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000492120.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 ENST00000492120.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006795.hg.1 NM_001621.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline