| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000927.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000927.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000927.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000927.hg.1 |
TC06000087.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006967.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147704.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147704.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147704.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147704.hg.1 |
ENST00000379375.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147704.hg.1 |
NM_001168319.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147704.hg.1 |
NM_001955.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147704.hg.1 |
TC0600006967.hg.1 |
Clariom D TC |