Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00186349_m1
          See other AGPS GE Assays ›
          Gene Symbol:
          AGPS
          Gene Name:
          alkylglycerone phosphate synthase
          Gene Aliases:
          ADAP-S, ADAS, ADHAPS, ADPS, ALDHPSY, RCDP3
          Chromosome Location:
          Chr.2: 177392743 - 177543836 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00186349_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AGPS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003659.3 NP_003650.1 1 - 2 310 135
          GenBank mRNA AK309714.1 - 1 - 2 409 135
          AK314259.1 - 1 - 2 280 135
          AY544121.1 - 1 - 2 263 135
          BC141820.1 - 1 - 2 275 40148148 135
          DC402978.1 - 1 - 2 312 135
          Y09443.1 - 1 - 2 278 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200010100.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001065.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AGPS

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid biosynthetic process
          ether lipid biosynthetic process
          oxidation-reduction process
          protein binding
          alkylglycerone-phosphate synthase activity
          oxidoreductase activity, acting on CH-OH group of donors
          FAD binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02015823.hg.1 TC02001065.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02015823.hg.1 ENST00000264167.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02015823.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02015823.hg.1 ENST00000536686.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02015823.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02015820.hg.1 TC02001065.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02015820.hg.1 ENST00000264167.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02015820.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02015820.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000460342.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000460342.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 ENST00000460342.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200170746.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170746.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200170746.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170746.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200170746.hg.1 TC0200010100.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200170751.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170751.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170751.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200170751.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170751.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170751.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200170751.hg.1 TC0200010100.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200170749.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170749.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170749.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200170749.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170749.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200170749.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200170749.hg.1 TC0200010100.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000264167.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000409888.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000460342.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000460342.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 ENST00000460342.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq
          Affymetrix Human Clariom D Assay TC0200010100.hg.1 NM_003659.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0